Skip to Content
Merck
All Photos(1)

Documents

601527

Sigma-Aldrich

Lithium-7Li hydroxide monohydrate

99.9 atom % 7Li

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
7LiOH·H2O
CAS Number:
Molecular Weight:
42.04
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.12

description

Special high CP

Quality Level

isotopic purity

99.9 atom % 7Li

form

solid

density

1.51 g/mL at 25 °C (lit.)

mass shift

depleted

SMILES string

[7Li+].O.[OH-]

InChI

1S/Li.2H2O/h;2*1H2/q+1;;/p-1/i1+0;;

InChI key

GLXDVVHUTZTUQK-GLJCYROLSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason C Wood et al.
Applied spectroscopy, 75(2), 199-207 (2020-08-09)
Laser-induced breakdown spectroscopy (LIBS) is a technique capable of portable, quantitative elemental analysis; however, quantitative isotopic determination of samples in situ has not yet been demonstrated. This research demonstrates the ability of LIBS to quantitatively determine concentrations of 6Li in
P Sivakumar et al.
Ultrasonics sonochemistry, 26, 332-339 (2015-03-10)
LiNi0.5Mn1.5O4 was synthesized as a cathode material for Li-ion batteries by a sonochemical reaction followed by annealing, and was characterized by XRD, SEM, HRTEM and Raman spectroscopy in conjunction with electrochemical measurements. Two samples were prepared by a sonochemical process
Martina Riesová et al.
Journal of chromatography. A, 1364, 276-288 (2014-09-13)
In this paper we determine acid dissociation constants, limiting ionic mobilities, complexation constants with β-cyclodextrin or heptakis(2,3,6-tri-O-methyl)-β-cyclodextrin, and mobilities of resulting complexes of profens, using capillary zone electrophoresis and affinity capillary electrophoresis. Complexation parameters are determined for both neutral and
James D Oliver et al.
Biotechnology and applied biochemistry, 62(3), 329-342 (2014-07-22)
Free-solution capillary electrophoresis (CE), or capillary zone electrophoresis, with direct UV detection was used for the first time for the determination of mono- and disaccharides, sugar alcohols, and ethanol in fermentation broths. Sample preparation proved to be minimal: no derivatization
Ludmila Müllerová et al.
Journal of chromatography. A, 1384, 147-154 (2015-02-11)
Interactions among analyte forms that undergo simultaneous dissociation/protonation and complexation with multiple selectors take the shape of a highly interconnected multi-equilibrium scheme. This makes it difficult to express the effective mobility of the analyte in these systems, which are often

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service