Merck
  • Pannexin-1-mediated ATP release from area CA3 drives mGlu5-dependent neuronal oscillations.
Pannexin-1-mediated ATP release from area CA3 drives mGlu5-dependent neuronal oscillations.
Neuropharmacology (2015-02-04)
Jan Lopatář, Nicholas Dale, Bruno G Frenguelli
ABSTRACT
The activation of Group I metabotropic glutamate receptors (GI mGluRs) in the hippocampus results in the appearance of persistent bursts of synchronised neuronal activity. In response to other stimuli, such activity is known to cause the release of the purines ATP and its neuroactive metabolite, adenosine. We have thus investigated the potential release and role of the purines during GI mGluR-induced oscillations in rat hippocampal areas CA3 and CA1 using pharmacological techniques and microelectrode biosensors for ATP and adenosine. The GI mGluR agonist DHPG induced both persistent oscillations in neuronal activity and the release of adenosine in areas CA1 and CA3. In contrast, the DHPG-induced release of ATP was only observed in area CA3. Whilst adenosine acting at adenosine A1 receptors suppressed DHPG-induced burst activity, the activation of mGlu5 and P2Y1 ATP receptors were necessary for the induction of DHPG-induced oscillations. Selective inhibition of pannexin-1 hemichannels with a low concentration of carbenoxolone (10 μM) or probenecid (1 mM) did not affect adenosine release in area CA3, but prevented both ATP release in area CA3 and DHPG-induced bursting. These data reveal key aspects of GI mGluR-dependent neuronal activity that are subject to bidirectional regulation by ATP and adenosine in the initiation and pacing of burst firing, respectively, and which have implications for the role of GI mGluRs in seizure activity and neurodevelopmental disorders.
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder
Sigma-Aldrich
6-Methyl-2-(phenylethynyl)pyridine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
2-Ethylphenol, 99%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Adenosine, suitable for cell culture, BioReagent
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, Vetec, reagent grade, 99%