Skip to Content
Merck
All Photos(2)

Key Documents

E44000

Sigma-Aldrich

2-Ethylphenol

99%

Synonym(s):

1-Ethyl-2-hydroxybenzene, 2-Ethylphenol, Phlorol, o-Ethylphenol, o-Hydroxyethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4OH
CAS Number:
Molecular Weight:
122.16
Beilstein:
1099397
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.536 (lit.)

bp

195-197 °C (lit.)

mp

−18 °C (lit.)

density

1.037 g/mL at 25 °C (lit.)

SMILES string

CCc1ccccc1O

InChI

1S/C8H10O/c1-2-7-5-3-4-6-8(7)9/h3-6,9H,2H2,1H3

InChI key

IXQGCWUGDFDQMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Lopatář et al.
Neuropharmacology, 93, 219-228 (2015-02-04)
The activation of Group I metabotropic glutamate receptors (GI mGluRs) in the hippocampus results in the appearance of persistent bursts of synchronised neuronal activity. In response to other stimuli, such activity is known to cause the release of the purines
Pasquale Filannino et al.
Applied and environmental microbiology, 80(24), 7574-7582 (2014-09-28)
The metabolism of hydroxycinnamic acids by strictly heterofermentative lactic acid bacteria (19 strains) was investigated as a potential alternative energy route. Lactobacillus curvatus PE5 was the most tolerant to hydroxycinnamic acids, followed by strains of Weissella spp., Lactobacillus brevis, Lactobacillus
T Schettgen et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 995-996, 93-100 (2015-06-06)
We have developed and validated a simple and sensitive method for the determination of urinary phenol as well as the urinary metabolites of toluene and ethylbenzene in one analytical run. After enzymatic hydrolysis for the cleavage of conjugates overnight, the
Cynthia D Selassie et al.
Journal of medicinal chemistry, 48(23), 7234-7242 (2005-11-11)
In this comprehensive study on the caspase-mediated apoptosis-inducing effect of 51 substituted phenols in a murine leukemia cell line (L1210), we determined the concentrations needed to induce caspase activity by 50% (I50) and utilized these data to develop the following

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service