Merck
  • Comparison of Aroma-Active Volatiles in Oolong Tea Infusions Using GC-Olfactometry, GC-FPD, and GC-MS.
Comparison of Aroma-Active Volatiles in Oolong Tea Infusions Using GC-Olfactometry, GC-FPD, and GC-MS.
Journal of agricultural and food chemistry (2015-08-11)
JianCai Zhu, Feng Chen, LingYing Wang, YunWei Niu, Dan Yu, Chang Shu, HeXing Chen, HongLin Wang, ZuoBing Xiao
ABSTRACT
The aroma profile of oolong tea infusions (Dongdingwulong, DDWL; Tieguanyin, TGY; Dahongpao, DHP) were investigated in this study. Gas chromatography-olfactometry (GC-O) with the method of aroma intensity (AI) was employed to investigate the aroma-active compounds in tea infusions. The results presented forty-three, forty-five, and forty-eight aroma-active compounds in the TGY, DHP, and DDWL infusions, including six, seven, and five sulfur compounds, respectively. In addition, the concentration of volatile compounds in the tea infusions was further quantitated by solid phase microextraction-gas chromatography (SPME)-GC-MS and SPME-GC-flame photometric detection (FPD). Totally, seventy-six and thirteen volatile and sulfur compounds were detected in three types of tea infusions, respectively. Quantitative results showed that forty-seven aroma compounds were at concentrations higher than their corresponding odor thresholds. On the basis of the odor activity values (OAVs), 2-methylpropanal (OAV: 230-455), 3-methylbutanal (1-353), 2-methylbutanal (34-68), nerolidol (108-184), (E)-2-heptenal (148-294), hexanal (134-230), octanal (28-131), β-damascenone (29-59), indole (96-138), 6-methyl-5-hepten-2-one (34-67), (R)-(-)-linalool (63-87), and dimethyl sulfide (7-1320) presented relatively higher OAVs than those of other compounds, indicating the importance of these compounds in the overall aroma of tea infusions.
Product Description
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, metal alloy fiber, for use with manual holder
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with manual holder
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), fused silica fiber, df 65 μm(PDMS/DVB, for use with manual holder, needle size 23 ga
Supelco
SPME Sampling Stand, for use with 15 mL vials
Sigma-Aldrich
Indole, ≥99%
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 24 ga
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with manual holder
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
2-Pentylfuran, ≥98%, FG
Sigma-Aldrich
Benzothiazole, 96%
Sigma-Aldrich
Geraniol, 98%
Sigma-Aldrich
trans,trans-2,4-Heptandienal, ≥88%
Sigma-Aldrich
Furan, ≥99%