Skip to Content
Merck
All Photos(1)

Documents

W316407

Sigma-Aldrich

trans,trans-2,4-Heptandienal

≥88%

Synonym(s):

2,4-Heptadienal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCH=CHCHO
CAS Number:
Molecular Weight:
110.15
FEMA Number:
3164
Beilstein:
1699244
EC Number:
Council of Europe no.:
729
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.084
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Halal
Kosher

Assay

≥88%

refractive index

n20/D 1.534 (lit.)

bp

84-84.5 °C (lit.)

density

0.881 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; green; oily; vegetable

SMILES string

CC\C=C\C=C\C=O

InChI

1S/C7H10O/c1-2-3-4-5-6-7-8/h3-7H,2H2,1H3/b4-3+,6-5+

InChI key

SATICYYAWWYRAM-VNKDHWASSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dandan Qi et al.
Food chemistry, 265, 189-199 (2018-06-10)
The effects of rapid aging technology on aroma compounds and characteristic of white tea were investigated in comparison with natural aged white tea (NAWT) and fresh white tea (FWT). Volatile compounds of white teas were extracted using headspace-solid phase microextraction
JianCai Zhu et al.
Food chemistry, 221, 1484-1490 (2016-12-17)
Twenty-four kinds of representative aroma compounds in Oolong tea were selected to evaluate the interactions by the variation in the threshold values of these compounds before, and after, they were mixed. Result demonstrated that the ratios of the referenced threshold
Nadia Ruocco et al.
Scientific reports, 9(1), 517-517 (2019-01-27)
Diatom-derived polyunsaturated aldehydes (PUAs), decadienal, heptadienal and octadienal, derive from the oxidation of fatty acids and have cytotoxic and anticancer effects. PUAs, tested separately, induce malformations in sea urchin Paracentrotus lividus embryos. Decadienal induces the worst malformations and lowest survival
Jon Alberdi-Cedeño et al.
Antioxidants (Basel, Switzerland), 9(6) (2020-06-25)
For the first time, an important number of oxylipins have been identified and quantified in corn oil submitted to mild oxidative conditions at each time of their oxidation process. This oil can be considered as a model system of edible
Serkan Selli et al.
Journal of the science of food and agriculture, 98(11), 4104-4111 (2018-02-02)
Olives are processed in different ways depending on consumption habits, which vary between countries. Different de-bittering methods affect the aroma and aroma-active compounds of table olives. This study focused on analyzing the aroma and aroma-active compounds of black dry-salted olives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service