Skip to Content
Merck
All Photos(2)

Key Documents

57302

Supelco

SPME fiber assembly Polydimethylsiloxane (PDMS)

greener alternative

df 7 μm(PDMS, needle size 24 ga, for use with manual holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854

material

fused silica fiber
plain green hub

Quality Level

needle size

24 ga

packaging

pkg of 3 ×  ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

7 μm (PDMS)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

PDMS coating

application(s)

food and beverages

compatibility

for analyte group volatiles (MW 80-500)
for use with manual holder

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Application

Nonpolar High Molecular Weight Compounds

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid Phase Microextraction -- A Solventless Sample Preparation Method for Organic Compounds in Water
Shirey, R., et al.
American Environmental Laboratory, 43-45 (1994)
Susan Sonchik Marine et al.
Journal of chromatographic science, 41(1), 31-35 (2003-02-25)
Limonene is a common component found in consumer goods ranging from beverages to cleaning compounds. Limonene oxidation products, however, have a less desirable flavor and fragrance. Early detection of limonene oxide formation would aid quality control. A method is developed
Analysis of Vitamin K in Green Tea Leafs and Infusions by SPME-GC-FID
Reto, M., et al.
Food Chemistry, 100, 405-411 (2006)
SPME, A New Tool in Pheromone Identification in Lipidoptera
Frerot, B., et al.
Journal of High Resolution Chromatography, 20(6), 340-342 (1997)
Liu et al.
Journal of chromatographic science, 38(9), 377-382 (2000-09-30)
An empirical model describing the relationship between the partition coefficients (K) of perfume materials in the solid-phase microextraction (SPME) fiber stationary phase and the Linearly Temperature Programmed Retention Index (LTPRI) is obtained. This is established using a mixture of eleven

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service