Merck
  • Simple measurements for prediction of drug release from polymer matrices - Solubility parameters and intrinsic viscosity.
Simple measurements for prediction of drug release from polymer matrices - Solubility parameters and intrinsic viscosity.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-02-11)
Claus G Madsen, Anders Skov, Stefania Baldursdottir, Thomas Rades, Lene Jorgensen, Natalie J Medlicott
ABSTRACT
This study describes how protein release from polymer matrices correlate with simple measurements on the intrinsic viscosity of the polymer solutions used for casting the matrices and calculations of the solubility parameters of polymers and solvents used. Matrices of poly(dl-lactide-co-glycolide) (PLGA) were cast with bovine serum albumin (BSA) as a model drug using different solvents (acetone, dichloromethane, ethanol and water). The amount of released protein from the different matrices was correlated with the Hildebrand and Hansen solubility parameters of the solvents, and the intrinsic viscosity of the polymer solutions. Matrix microstructure was investigated by transmission and scanning electron microscopy (TEM and SEM). Polycaprolactone (PCL) matrices were used in a similar way to support the results for PLGA matrices. The maximum amount of BSA released and the release profile from PLGA matrices varied depending on the solvent used for casting. The maximum amount of released BSA decreased with higher intrinsic viscosity, and increased with solubility parameter difference between the solvent and polymer used. The solvent used also had an effect on the matrix microstructure as determined by TEM and SEM. Similar results were obtained for the PCL polymer systems. The smaller the difference in the solubility parameter between the polymer and the solvent used for casting a polymer matrix, the lower will be the maximum protein release. This is because of the presence of smaller pore sizes in the cast matrix if a solvent with a solubility parameter close to the one of the polymer is used. Likewise, the intrinsic viscosity of the polymer solution increases as solubility parameter differences decrease, thus, simple measurements of intrinsic viscosity and solubility parameter difference, allow the prediction of protein release profiles.
Sigma-Aldrich
Bovine Serum Albumin, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Protein standard, Micro Standard, liquid
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Albumin from sheep serum, lyophilized powder
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, Protease, essentially free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially γ-globulin free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin solution, 35% in DPBS, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Protein standard, liquid, 2 mg protein/ml
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥97% (agarose gel electrophoresis), 1X Crystallized
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, microbiologically tested, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, For use as a marker in SDS-PAGE
Sigma-Aldrich
Bovine Serum Albumin, dimer mol wt ~132 kDa
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)