Skip to Content
Merck
All Photos(4)

Key Documents

A1470

Sigma-Aldrich

Bovine Serum Albumin

lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27
Pricing and availability is not currently available.

biological source

bovine

product line

BioReagent

Assay

≥98% (agarose gel electrophoresis)

form

lyophilized powder

mol wt

~66 kDa

purified by

heat shock fractionation

packaging

poly bottle of

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤20 ppm Pb
≤3 EU/mg endotoxin

pH

7

solubility

water: soluble (40 mg/ml)

UniProt accession no.

foreign activity

standard plate count (Bioburden) <500 (CFU/g)

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Features and Benefits

  • Tested for use in cell culture
  • Low endotoxin

Preparation Note

Manufactured by a novel process that maintains the lipid and growth factor characteristics of cold-ethanol precipitated albumins.
Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.

Legal Information

Cohn Analog is a trademark of Proliant, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akshay Shekhar et al.
Scientific reports, 8(1), 9944-9944 (2018-07-04)
Rapid impulse propagation is a defining attribute of the pectinated atrial myocardium and His-Purkinje system (HPS) that safeguards against atrial and ventricular arrhythmias, conduction block, and myocardial dyssynchrony. The complex transcriptional circuitry that dictates rapid conduction remains incompletely understood. Here
Holger Bronger et al.
Oncology letters, 13(6), 4224-4230 (2017-06-11)
Cathepsin B (CTSB) is a lysosomal cysteine protease that has been linked to the progression of breast cancer, for example by activation of other proteases and tumor-promoting cytokines, thereby supporting tumor invasion and metastasis. Previously, it was shown that CTSB
Shinichiro Atsumi et al.
Communications biology, 3(1), 588-588 (2020-10-18)
Despite the accumulating evidences of the significance of humoral cancer immunity, its molecular mechanisms have largely remained elusive. Here we show that B-cell repertoire sequencing of 102 clinical gastric cancers and molecular biological analyses unexpectedly reveal that the major humoral
Marcos V A S Navarro et al.
PLoS biology, 9(2), e1000588-e1000588 (2011-02-10)
The bacterial second messenger bis-(3'-5') cyclic dimeric guanosine monophosphate (c-di-GMP) has emerged as a central regulator for biofilm formation. Increased cellular c-di-GMP levels lead to stable cell attachment, which in Pseudomonas fluorescens requires the transmembrane receptor LapD. LapD exhibits a
Manoj K Pandey et al.
Molecular pharmacology, 75(3), 525-533 (2008-12-24)
The aim of the current study is to determine whether butein (3,4,2',4'-tetrahydroxychalcone) exhibits antiproliferative effects against tumor cells through suppression of the signal transducer and activator of transcription 3 (STAT3) activation pathway. We investigated the effects of butein on constitutive

Articles

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Related Content

Learn about bovine serum albumin (BSA) structure, molecular weight, function, solubility, how BSA is made, purification methods, applications and purchasing.

Learn about bovine serum albumin (BSA) structure, molecular weight, function, solubility, how BSA is made, purification methods, applications and purchasing.

Learn about bovine serum albumin (BSA) structure, molecular weight, function, solubility, how BSA is made, purification methods, applications and purchasing.

Learn about bovine serum albumin (BSA) structure, molecular weight, function, solubility, how BSA is made, purification methods, applications and purchasing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service