Merck
  • Reactivity of an All-Ferrous Iron-Nitrogen Heterocubane under Reductive and Oxidative Conditions.
Reactivity of an All-Ferrous Iron-Nitrogen Heterocubane under Reductive and Oxidative Conditions.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-09-17)
Crispin Lichtenberg, Demyan E Prokopchuk, Mario Adelhardt, Liliana Viciu, Karsten Meyer, Hansjörg Grützmacher
ABSTRACT
The reactivity of the all-ferrous FeN heterocubane [Fe4 (Ntrop)4 ] (1) with i) Brønsted acids, ii) σ-donors, iii) σ-donors/π-acceptors, and iv) one-electron oxidants has been investigated (trop = 5H-dibenzo[a,d]cyclo-hepten-5-yl). 1 showed self-re-assembling after reactions with i) and proved surprisingly inert in reactions with ii) and iii), with the exception of CO. Reductive and oxidative cluster degradation was observed in reactions with CO and TEMPO, respectively. These reactions yielded new cluster compounds, namely a trinuclear bis(μ3 -imido) 48 electron complex in the former case and a tetranuclear all ferric μ-oxo-μ-imido species in the latter case. Characterization techniques include NMR and in situ IR spectroscopy, single crystal X-ray analysis, Mössbauer spectroscopy, cyclic voltammetry, magnetic susceptibility measurements, and DFT calculations.
Sigma-Aldrich
Trimethylphosphine solution, 1.0 M in THF
Sigma-Aldrich
Trimethylphosphine solution, 1.0 M in toluene
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
TEMPO, 98%
Sigma-Aldrich
Trimethylphosphine, 97%
Sigma-Aldrich
TEMPO, purified by sublimation, 99%