Merck
  • Moraxella catarrhalis induces an immune response in the murine lung that is independent of human CEACAM5 expression and long-term smoke exposure.
Moraxella catarrhalis induces an immune response in the murine lung that is independent of human CEACAM5 expression and long-term smoke exposure.
American journal of physiology. Lung cellular and molecular physiology (2015-06-07)
Birgitt Gutbier, Katja Fischer, Jan-Moritz Doehn, Carolin von Lachner, Christian Herr, Esther Klaile, Ursula Frischmann, Bernhard B Singer, Kristian Riesbeck, Wolfgang Zimmermann, Norbert Suttorp, Sebastian Bachmann, Robert Bals, Martin Witzenrath, Hortense Slevogt
ABSTRACT
In patients with chronic obstructive pulmonary disease (COPD), Moraxella catarrhalis infection of the lower airways is associated with chronic colonization and inflammation during stable disease and acute exacerbations. Chronic smoke exposure induces chronic inflammation and impairs mucociliary clearance, thus contributing to bacterial colonization of the lower airways in COPD patients. The human-specific carcinoembryonic antigen-related cell adhesion molecule (CEACAM) 5, expressed in human airways, has been shown to contribute to epithelial colonization of CEACAM-binding pathogens. To investigate the impact of CEACAM5 expression on pulmonary M. catarrhalis colonization, we infected mice transgenic for human CEACAM5 (hCEACAM5) and wild type mice intratracheally with M. catarrhalis with or without preceding smoke exposure and analyzed bacterial colonization and local and systemic inflammation. Our results show that airway infection with M. catarrhalis accelerated acute local but not systemic inflammation, albeit independent of hCEACAM5 expression. Long-term smoke exposure alone or prior to M. catarrhalis infection did not contribute to increased local or systemic inflammation. No difference was found in pulmonary clearance of M. catarrhalis in hCEACAM5-transgenic mice compared with wild-type mice. Smoke exposure neither altered time nor extent of persistence of M. catarrhalis in the lungs of both genotypes. In conclusion, M. catarrhalis induced a local acute immune response in murine airways. Neither hCEACAM5 expression nor chronic smoke exposure nor a combination of both was sufficient as prerequisites for the establishment of chronic M. catarrhalis colonization. Our results demonstrate the difficulties in mirroring conditions of chronic airways colonization of M. catarrhalis in a murine model.
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sigma-Aldrich
Thrombopoietin from mouse, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture, >97% (SDS-PAGE)
Sigma-Aldrich
Xylazine, ≥99%
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Carbon nanofibers, graphitized, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Carbon nanofibers, pyrolitically stripped, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Thrombopoietin human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.9 atom % 12C
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis