Merck
  • Identification of N-linked glycosylation and putative O-fucosylation, C-mannosylation sites in plasma derived ADAMTS13.
Identification of N-linked glycosylation and putative O-fucosylation, C-mannosylation sites in plasma derived ADAMTS13.
Journal of thrombosis and haemostasis : JTH (2014-07-01)
N Sorvillo, P H Kaijen, M Matsumoto, Y Fujimura, C van der Zwaan, F C Verbij, W Pos, R Fijnheer, J Voorberg, A B Meijer
ABSTRACT
Acquired deficiency of ADAMTS13 causes a rare and life-threatening disorder called thrombotic thrombocytopenic purpura (TTP). Several studies have shown that aberrant glycosylation can play an important role in the pathogenesis of autoimmune diseases.N-linked glycosylation and putative O-fucosylation sites have been predicted or identified in recombinant ADAMTS13. However, it is not known which of these sites are glycosylated in plasma derived ADAMTS13. Here we investigated the presence of putative O-fucosylation, C-mannosylation and N-linked glycosylation sites on plasma derived ADAMTS13. Sites of N-linked glycosylation were determined by the use of peptide N-glycosidase-F (PNGase F), which removes the entire carbohydrate from the side chain of asparagines. Nine of the 10 predicted N-linked glycosylation sites were identified in or near the metalloproteinase,spacer, thrombospondin type 1 repeat (TSR1) and the CUB domain of plasma ADAMTS13. Moreover, six putative O-fucosylated sites were identified in the TSR domains of plasma ADAMTS13 by performing searches of the tandem mass spectrometry (MS/MS) data for loss of hexose (162 Da), deoxyhexose (146 Da), or hexose deoxyhexose(308 Da). The use of electron transfer dissociation (ETD) allowed for unambiguous identification of the modified sites. In addition to putative O-fucosylation and N-linked glycosylation, two putative C-mannosylation sites were identified within the TSR1 and TSR4 domains of ADAMTS13. Our data identify several glycosylation sites on plasma derived ADAMTS13. We anticipate that our findings may be relevant for the initiation of autoimmune reactivity against ADAMTS13 in patients with acquired TTP.
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
SAFC
BIS-TRIS
SAFC
BIS-TRIS
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)