Skip to Content
Merck
All Photos(3)

Documents

B7535

Sigma-Aldrich

BIS-TRIS

BioXtra, ≥98.0% (titration)

Synonym(s):

2,2-Bis(hydroxymethyl)-2,2′,2″-nitrilotriethanol, 2-Bis(2-hydroxyethyl)amino-2-(hydroxymethyl)-1,3-propanediol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H19NO5
CAS Number:
Molecular Weight:
209.24
Beilstein:
2205275
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Assay

≥98.0% (titration)

form

powder

impurities

≤0.005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

pH

9.0-10.1 (20-25 °C, 1 M in H2O)

useful pH range

5.8-7.2

pKa (25 °C)

6.5

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

application(s)

diagnostic assay manufacturing

SMILES string

OCCN(CCO)C(CO)(CO)CO

InChI

1S/C8H19NO5/c10-3-1-9(2-4-11)8(5-12,6-13)7-14/h10-14H,1-7H2

InChI key

OWMVSZAMULFTJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Optimization of Exosome Isolation: Research on optimizing exosome isolation emphasized the utility of BIS-TRIS in establishing buffer systems that enhance the accuracy of exosome size and morphology analysis, critical for diagnostic applications (Ishii et al., 2023).

Other Notes

Easily compare specifications for BIS-TRIS products with the BIS-TRIS specification table.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth M MacDonald et al.
Disease models & mechanisms, 7(4), 471-481 (2014-02-08)
The purpose of our study was to compare two acquired muscle atrophies and the use of myostatin inhibition for their treatment. Myostatin naturally inhibits skeletal muscle growth by binding to ActRIIB, a receptor on the cell surface of myofibers. Because
Lesley Cheng et al.
Kidney international, 86(2), 433-444 (2013-12-20)
Micro RNAs (miRNAs) have been shown to circulate in biological fluids and are enclosed in vesicles such as exosomes; they are present in urine and represent a noninvasive methodology to detect biomarkers for diagnostic testing. The low abundance of RNA
Alastair W Skeffington et al.
Plant physiology, 165(2), 866-879 (2014-05-02)
The first step on the pathway of starch degradation in Arabidopsis (Arabidopsis thaliana) leaves at night is the phosphorylation of starch polymers, catalyzed by glucan, water dikinase (GWD). It has been suggested that GWD is important for the control of
Zahra Sabouri et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(25), E2567-E2575 (2014-05-14)
The best-understood mechanisms for achieving antibody self/non-self discrimination discard self-reactive antibodies before they can be tested for binding microbial antigens, potentially creating holes in the repertoire. Here we provide evidence for a complementary mechanism: retaining autoantibodies in the repertoire displayed
X Chen et al.
Oncogene, 33(39), 4724-4734 (2013-10-22)
Uveal melanoma (UM) is a genetically and biologically distinct type of melanoma, and once metastatic there is no effective treatment currently available. Eighty percent of UMs harbor mutations in the Gαq family members GNAQ and GNA11. Understanding the effector pathways

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service