Merck
  • A monomeric [Zr(CO3)4](4-) complex in an ammonium zirconium carbonate aqueous solution studied by extended X-ray absorption fine structure, Raman and nuclear magnetic resonance spectroscopy.
A monomeric [Zr(CO3)4](4-) complex in an ammonium zirconium carbonate aqueous solution studied by extended X-ray absorption fine structure, Raman and nuclear magnetic resonance spectroscopy.
Dalton transactions (Cambridge, England : 2003) (2014-11-12)
Fumiyuki Takasaki, Kazuhiko Fujiwara, Yasushi Nakajima, Taku Nishikawa, Hyuma Masu, Mamoru Imanari, Yuki Hidaka, Nobuaki Ogawa
ABSTRACT
The speciation of zirconium complexes in ammonium zirconium carbonate (AZC) solutions should be further studied in order to develop high-performance AZC complexes for use in elaborate industrial materials. Thus, the speciation and the structure of various zirconium species in ammonium zirconium carbonate solutions were investigated over a wide range of total carbonate and ammonium concentrations. By employing extended X-ray absorption fine structure analysis, and Raman and (13)C NMR spectroscopy, it was found that four carbonate ions coordinate bidentately to the zirconium ion. In addition, the exact concentration ranges of carbonate and ammonium in AZC necessary to generate the monomeric zirconium carbonate complex [Zr(CO3)4](4-) were determined.
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Boron nitride, powder, ~1 μm, 98%
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
Sigma-Aldrich
Boron nitride, nanopowder, <150 nm avg. part. size (TEM), 99% trace metals basis
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Potassium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
USP
Potassium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Potassium bicarbonate, ACS reagent, 99.7%, powder, crystals or granules
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol
Sigma-Aldrich
Potassium bicarbonate, ≥99.95% trace metals basis, 99.7-100.5% dry basis