Merck
  • Analysis of changes in SUMO-2/3 modification during breast cancer progression and metastasis.
Analysis of changes in SUMO-2/3 modification during breast cancer progression and metastasis.
Journal of proteome research (2014-07-30)
Divya Subramonian, Sarita Raghunayakula, Jesper V Olsen, Karen A Beningo, Wulf Paschen, Xiang-Dong Zhang
ABSTRACT
SUMOylation is an essential posttranslational modification and regulates many cellular processes. Dysregulation of SUMOylation plays a critical role in metastasis, yet how its perturbation affects this lethal process of cancer is not well understood. We found that SUMO-2/3 modification is greatly up-regulated in metastatic breast cancer cells compared with nonmetastatic control cells. To identify proteins differentially modified by SUMO-2/3 between metastatic and nonmetastatic cells, we established a method in which endogenous SUMO-2/3 conjugates are labeled by stable isotope labeling by amino acids in cell culture (SILAC), immunopurified by SUMO-2/3 monoclonal antibodies and epitope-peptide elution, and analyzed by quantitative mass spectrometry. We identified 66 putative SUMO-2/3-conjugated proteins, of which 15 proteins show a significant increase/decrease in SUMO-2/3 modification in metastatic cells. Targets with altered SUMOylation are involved in cell cycle, migration, inflammation, glycolysis, gene expression, and SUMO/ubiquitin pathways, suggesting that perturbations of SUMO-2/3 modification might contribute to metastasis by affecting these processes. Consistent with this, up-regulation of PML SUMO-2/3 modification corresponds to an increased number of PML nuclear bodies (PML-NBs) in metastatic cells, whereas up-regulation of global SUMO-2/3 modification promotes 3D cell migration. Our findings provide a foundation for further investigating the effects of SUMOylation on breast cancer progression and metastasis.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Pricing and availability is not currently available.
Supelco
Acetonitrile, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Suberic acid bis(N-hydroxysuccinimide ester), ≥95%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Ultrapure Acetonitrile
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Pricing and availability is not currently available.
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Glutathione, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.