Merck
  • Immunomodulatory effect of diethylcarbamazine in mice infected with Nocardia brasiliensis.
Immunomodulatory effect of diethylcarbamazine in mice infected with Nocardia brasiliensis.
International immunopharmacology (2014-08-26)
M García-Hernández, M A Castro-Corona, J C Segoviano-Ramírez, N W Brattig, C E Medina-De la Garza
ABSTRACT
We tested whether diethylcarbamazine (DEC) or ivermectin (IVM), both antiparasitic drugs with reported immunomodulatory properties, were able to affect the immune system to potentiate host defense mechanisms and protect against actinomycetoma in a mouse model. Male BALB/c mice of 10-12 weeks of age were injected with either Nocardia brasiliensis or saline solution. Recorded were the effects of a treatment by DEC (6 mg/kg per os daily for one week) or IVM (200 μg/kg subcutaneously on days 1 and 3) on (i) the development of mycetoma lesion, (ii) the expression of reactive oxygen intermediates (ROI) by phagocytes, (iii) the proliferation index of lymphocytes and (iv) antibody production of IgG and IgM. After an initial lesion in all mice, DEC inhibited a full development and progression of actinomycetoma resulting in a reduced lesion size (p < 0.001). IVM had no inhibitory effect on the development of mycetoma. Furthermore, DEC treatment was associated with a significant enhancement of ROI expression (p < 0.05) by polymorphonuclear neutrophils at day 3 after infection. Lymphocyte proliferation in response to N. brasiliensis antigens and concanavalin A in DEC-treated group was higher than in non-treated group at day 21 and 28 postinfection (p < 0.01). Significant changes in antibody response were not observed. By all parameters tested, DEC was superior to IVM regarding immunostimulatory potency. In conclusion, DEC expressed an in vivo influence on the immune status during the infection by N. brasiliensis leading to retrogression of the mycetoma and increasing cellular immune responses. Our findings may indicate a potential use of DEC as a putative adjuvant in infectious disease or vaccination.
Product Description
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Diethylcarbamazine citrate, European Pharmacopoeia (EP) Reference Standard
USP
Diethylcarbamazine citrate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%