Skip to Content
Merck
All Photos(1)

Key Documents

PHR1553

Supelco

N,N-Dimethylformamide

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

N,N-Dimethylformamide, DMF, NSC 5356

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Product Name

Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1601500

vapor density

2.5 (vs air)

vapor pressure

2.7 mmHg ( 20 °C)

CofA

current certificate can be downloaded

autoignition temp.

833 °F

expl. lim.

15.2 %

packaging

ampule of 3x1.2 mL each

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.430 (lit.)

bp

153 °C (lit.)

mp

−61 °C (lit.)

density

0.944 g/mL (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Dimethylformamide is a polar solvent commonly used in organic synthesis, isolation of chlorophyll from plant tissues, etc. It also acts as a multipurpose precursor for formylation, amination, amino carbonylation, amidation and cyanation reactions.

Application

Solvent for many hydrophobic organic compounds.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2887 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Ollier et al.
Journal of dairy science, 97(8), 4942-4954 (2014-06-03)
A cow's risk of acquiring a new intramammary infection during the dry period increases with milk production at drying off and decreases as mammary gland involution progresses. A method commonly used to reduce milk production is a drastic reduction in
Letian Dou et al.
Nature communications, 5, 5404-5404 (2014-11-21)
Photodetectors capture optical signals with a wide range of incident photon flux density and convert them to electrical signals instantaneously. They have many important applications including imaging, optical communication, remote control, chemical/biological sensing and so on. Currently, GaN, Si and
Jun Nishida et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(52), 18442-18447 (2014-12-17)
The structural elasticity of metal-organic frameworks (MOFs) is a key property for their functionality. Here, we show that 2D IR spectroscopy with pulse-shaping techniques can probe the ultrafast structural fluctuations of MOFs. 2D IR data, obtained from a vibrational probe
L Piazza et al.
Nature communications, 6, 6407-6407 (2015-03-03)
Surface plasmon polaritons can confine electromagnetic fields in subwavelength spaces and are of interest for photonics, optical data storage devices and biosensing applications. In analogy to photons, they exhibit wave-particle duality, whose different aspects have recently been observed in separate
Guillaume E Beranger et al.
Endocrinology, 155(4), 1340-1352 (2014-02-11)
Osteoporosis and overweight/obesity constitute major worldwide public health burdens that are associated with aging. A high proportion of women develop osteoporosis and increased intraabdominal adiposity after menopause. which leads to bone fractures and metabolic disorders. There is no efficient treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service