Skip to Content
Merck
All Photos(1)

Key Documents

494488

Sigma-Aldrich

N,N-Dimethylformamide

biotech. grade, ≥99.9%

Synonym(s):

DMF, NSC 5356

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.21
grade:
biotech. grade
Assay:
≥99.9%
application(s):
peptide synthesis
bp:
153 °C (lit.)
vapor pressure:
2.7 mmHg ( 20 °C)

grade

biotech. grade

Quality Level

vapor density

2.5 (vs air)

vapor pressure

2.7 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

833 °F

expl. lim.

15.2 %

impurities

≤0.0005 meq/g Titr. acid
≤0.003 meq/g Titr. base
≤0.005% (water)
<8 ppm Free amines as dimethylamine

evapn. residue

<0.0005%

refractive index

n20/D 1.430 (lit.)

pH

7 (20 °C, 200 g/L)

bp

153 °C (lit.)

mp

−61 °C (lit.)

solubility

water: miscible

density

0.944 g/mL (lit.)

UV absorption

λ: 270 nm Amax: ≤1.0
λ: 275 nm Amax: ≤0.30
λ: 295 nm Amax: ≤0.10
λ: 310 nm Amax: ≤0.05
λ: 340 nm Amax: ≤0.01
λ: 400 nm Amax: ≤0.01

application(s)

peptide synthesis

storage temp.

room temp

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylformamide (DMF) is a polar organic solvent with a wide range of industrial applications. It is a precursor for numerous reactions such as formylation, aminocarbonylation, amination, amidation and cyanation. Along with phosphorus oxychloride, it forms Vilsmeier reagent used in the formylation of reactive aromatic and heteroaromatic substrates. Dielectric relaxation experiments on DMF have been conducted between 238.15K and 338.15K. The thermophysical properties of the molecular interactions between DMF and the ionic liquids, ammonium salts and imidazolium salts have been studied. The characteristics of solute-solvent interactions between ammonium nitrate and DMF have been investigated.

Application

N,N-dimethylformamide (DMF) has been used for the preparation of tetraformylcavitand precursor for generating octahedral nanocontainer.
Solvent for many hydrophobic organic compounds.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple one-pot multicomponent synthesis of an octahedral nanocontainer molecule.
Liu X and Warmuth R.
Nature Protocols, 2(5), 1288-1296 (2007)
N,N-Dimethylformamide: much more than a solvent.
Muzart J.
Tetrahedron, 65(40), 8313-8323 (2009)
Review article on Vilsmeier Haack reaction and its Applications.
Beniwal M and Jain N.
European Journal of Biomedical AND Pharmaceutical sciences, 2(3), 1340-1374 (2015)
Volumetric properties of ammonium nitrate in N,N-dimethylformamide.
Vranes M, et al.
The Journal of Chemical Thermodynamics, 54, 245-249 (2012)
Measurements and molecular interactions for N,N-dimethylformamide with ionic liquid mixed solvents.
Attri P, et al.
The Journal of Physical Chemistry B, 114(18), 6126-6133 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service