Merck
  • Macro/Nano-gel composite as an injectable and bioactive bulking material for the treatment of urinary incontinence.
Macro/Nano-gel composite as an injectable and bioactive bulking material for the treatment of urinary incontinence.
Biomacromolecules (2014-04-18)
Kyung Min Park, Joo Young Son, Jong Hoon Choi, In Gul Kim, Yunki Lee, Ji Youl Lee, Ki Dong Park
ABSTRACT
Many women around the world are suffering from urinary incontinence, defined as the unintentional leakage of urine by external abnormal pressure. Although various kinds of materials have been utilized to treat this disease, therapies that are more effective are still needed for the treatment of urinary incontinence. Here, we present a macro/nanogel composed of in situ forming gelatin-based macrogels and self-assembled heparin-based nanogels, which can serve as an injectable and bioactive bulking material for the treatment of urinary incontinence. The hybrid hydrogels were prepared via enzymatic reaction in the presence of horseradish peroxidase and hydrogen peroxide. Incorporating a growth factor (GF)-loaded heparin nanogel into a gelatin gel matrix enabled the hybrid gel matrix to release GF continuously up to 28 days. Moreover, we demonstrated that the hydrogel composites stimulated the regeneration of the urethral muscle tissue surrounding the urethral wall and promoted the recovery of their biological function when injected in vivo. Thus, the macro/nanohydrogels may provide an advanced therapeutic technique for the treatment of urinary incontinence as well as an application for regenerative medicine.
Product Description
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, purum, ≥99% (GC)
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Supelco
Triethylamine, analytical standard
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
Sigma-Aldrich
Succinic anhydride, ≥99% (GC)
Sigma-Aldrich
Tyramine, ≥98.0%
Sigma-Aldrich
Human bFGF ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Mouse bFGF ELISA Kit, for serum, plasma and cell culture supernatant