Skip to Content
Merck
All Photos(1)

Key Documents

1707894

USP

Valacyclovir Related Compound G

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-(Dimethylamino)pyridine, N,N-Dimethylpyridin-4-amine, DMAP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
Beilstein:
110354
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

valacyclovir

manufacturer/tradename

USP

mp

108-110 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN(C)c1ccncc1

InChI

1S/C7H10N2/c1-9(2)7-3-5-8-6-4-7/h3-6H,1-2H3

InChI key

VHYFNPMBLIVWCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Valacyclovir Related Compound G, also called as 4-(Dimethylamino)pyridine (DMAP) is an excellent catalyst for acylation of hindered alcohols and in chemical transformations. It is highly nucleophilic in nature.[1]

Application

A highly efficient catalyst for acylation reactions
Valacyclovir Related Compound G USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Valacyclovir Hydrochloride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 1

Target Organs

Nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

255.2 °F

Flash Point(C)

124 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Configurationally Stable Biaryl Analogues of 4-(Dimethylamino) pyridine: A Novel Class of Chiral Nucleophilic Catalysts.
Spivey, Alan C., et al.
The Journal of Organic Chemistry, 64.26, 9430-9443 (1999)
Masahiro Murakami et al.
Acta biomaterialia, 10(11), 4723-4729 (2014-07-20)
The objective of this study is to design a drug delivery system (DDS) for the in vivo promotion of macrophage recruitment. As the drug, a water-insoluble agonist of sphingosine-1-phosphate type 1 receptor (SEW2871) was selected. SEW2871 (SEW) was water-solubilized by
Bryan Hoang et al.
International journal of pharmaceutics, 471(1-2), 224-233 (2014-05-24)
Docetaxel (DTX) remains the only effective drug for prolonging survival and improving quality of life of metastatic castration resistant prostate cancer (mCRPC) patients. Despite some clinical successes with DTX-based therapies, advent of cumulative toxicity and development of drug resistance limit
Aniruddha Roy et al.
Molecular pharmaceutics, 11(8), 2592-2599 (2014-02-26)
Taxanes are a class of anticancer agents with a broad spectrum and have been widely used to treat a variety of cancer. However, its long-term use has been hampered by accumulating toxicity and development of drug resistance. The most extensively
Junya Ito et al.
Journal of chromatography. A, 1386, 53-61 (2015-02-18)
Increasing evidence suggests that phospholipid peroxidation plays important roles in the pathogenesis of various diseases, such as atherosclerosis. With regard to the biochemical processes that initiate phospholipid peroxidation in vivo, enzymatic conversion of phosphatidylcholine to phosphatidylcholine hydroperoxide (PCOOH) by lipoxygenase

Articles

Thin-layer chromatography method for detecting organic impurities in valacyclovir, an antiviral medication, following USP monograph guidelines.

Thin-layer chromatography method for detecting organic impurities in valacyclovir, an antiviral medication, following USP monograph guidelines.

Thin-layer chromatography method for detecting organic impurities in valacyclovir, an antiviral medication, following USP monograph guidelines.

Thin-layer chromatography method for detecting organic impurities in valacyclovir, an antiviral medication, following USP monograph guidelines.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service