Skip to Content
Merck
All Photos(1)

Key Documents

95294

Sigma-Aldrich

Hydrogen peroxide solution

tested according to Ph. Eur.

Synonym(s):

Hydrogenii peroxidum 30 per centum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

vapor pressure

23.3 mmHg ( 30 °C)

Assay

29.0-31.0% (M/M)

form

solution

shelf life

~2 yr

reaction suitability

reagent type: oxidant

density

1.110 g/cm3

storage temp.

2-8°C

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrogen peroxide is an reactive oxygen species formed as by-product of normal aerobic metabolism and increases under stress conditions.[1] Fluorometric method for the ultra-trace quantitation of hydrogen peroxide in the range of 10-11mole/ml has been described.[2] Hydrogen peroxide is formed as terminal product of the hydroperoxy radical in photochemical reactions. It plays role of an oxidant in heterogeneous processes. Trace level determination of hydrogen peroxide using the oxo[5,10,15,20- tetra(4-pyridyl)porphyrinato]titanium( IV) complex has been reported. [3]

Application

Hydrogen peroxide solution may be used for the epoxidation of lower olefins, catalyzed by titanium silicalite (TS-1).[3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Oxo [5, 10, 15, 20-tetra (4-pyridyl) porphyrinato] titanium (IV): an ultra-high sensitivity spectrophotometric reagent for hydrogen peroxide.
Matsubara C, et al.
Analyst, 117(11), 1781-1784 (1992)
THE FLUOROMETRIC ANALYSIS OF ULTRAMICRO QUANTITIES OF HYDROGEN PEROXIDE.
A S KESTON et al.
Analytical biochemistry, 11, 1-5 (1965-04-01)
Spectrofluorimetric determination of hydrogen peroxide scavenging activity.
Pazdzioch-Czochra M and Widenska A.
Analytica Chimica Acta, 452(2), 177-184 (2002)
Shoichi Iriguchi et al.
Blood, 125(2), 370-382 (2014-10-29)
Although overexpression of T-bet, a master transcription factor in type-1 helper T lymphocytes, has been reported in several hematologic and immune diseases, its role in their pathogenesis is not fully understood. In the present study, we used transgenic model mice
James R Van Brocklyn et al.
Cancer research, 74(19), 5364-5370 (2014-08-12)
Glioblastoma remains a devastating disease for which novel therapies are urgently needed. Here, we report that the Aurora-A kinase inhibitor alisertib exhibits potent efficacy against glioblastoma neurosphere tumor stem-like cells in vitro and in vivo. Many glioblastoma neurosphere cells treated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service