Merck
  • Utility of cerebrospinal fluid drug concentration as a surrogate for unbound brain concentration in nonhuman primates.
Utility of cerebrospinal fluid drug concentration as a surrogate for unbound brain concentration in nonhuman primates.
Drug metabolism and pharmacokinetics (2014-05-09)
Yoko Nagaya, Yoshitane Nozaki, Kazumasa Kobayashi, Osamu Takenaka, Yosuke Nakatani, Kazutomi Kusano, Tsutomu Yoshimura, Hiroyuki Kusuhara
ABSTRACT
In central nervous system drug discovery, cerebrospinal fluid (CSF) drug concentration (C(CSF)) has been widely used as a surrogate for unbound brain concentrations (C(u,brain)). However, previous rodent studies demonstrated that when drugs undergo active efflux by transporters, such as P-glycoprotein (P-gp), at the blood-brain barrier, the C(CSF) overestimates the corresponding C(u,brain). To investigate the utility of C(CSF) as a surrogate for interstitial fluid (ISF) concentration (C(ISF)) in nonhuman primates, this study simultaneously determined the C(CSF) and C(ISF) of 12 compounds, including P-gp substrates, under steady-state conditions in cynomolgus monkeys using intracerebral microdialysis coupled with cisternal CSF sampling. Unbound plasma concentrations of non- or weak P-gp substrates were within 2.2-fold of the C(ISF) or C(CSF), whereas typical P-gp substrates (risperidone, verapamil, desloratadine, and quinidine) showed ISF-to-plasma unbound (K(p,uu,ISF)) and CSF-to-plasma unbound concentration ratios (K(p,uu,CSF)) that were appreciably lower than unity. Although the K(p,uu,CSF) of quinidine, verapamil, and desloratadine showed a trend of overestimating the K(p,uu,ISF), K(p,uu,CSF) showed a good agreement with K(p,uu,ISF) within 3-fold variations for all compounds examined. C(u,brain) of some basic compounds, as determined using brain homogenates, overestimated the C(ISF) and C(CSF). Therefore, C(CSF) could be used as a surrogate for C(ISF) in nonhuman primates.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Risperidone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Antipyrine, analytical standard
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
Quinidine, crystallized, ≥98.0% (dried material, NT)
Ondansetron hydrochloride dihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ondansetron hydrochloride dihydrate, ≥98% (HPLC), powder
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
(±)-Propranolol hydrochloride, analytical standard
USP
Verapamil hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Quinidine, anhydrous
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Metoclopramide hydrochloride, solid
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
USP
Risperidone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
SAFC
Sodium chloride solution, 5 M
SAFC
HEPES
Supelco
Propranolol hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Desloratadine, powder, ≥98% (HPLC)
Sigma-Aldrich
Risperidone, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
USP
Ondansetron hydrochloride, United States Pharmacopeia (USP) Reference Standard