Skip to Content
Merck
All Photos(2)

Key Documents

M0763

Sigma-Aldrich

Metoclopramide hydrochloride

solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22ClN3O2 · HCl
CAS Number:
Molecular Weight:
336.26
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

solubility

H2O: soluble

originator

Baxter

SMILES string

Cl.CCN(CC)CCNC(=O)c1cc(Cl)c(N)cc1OC

InChI

1S/C14H22ClN3O2.ClH/c1-4-18(5-2)7-6-17-14(19)10-8-11(15)12(16)9-13(10)20-3;/h8-9H,4-7,16H2,1-3H3,(H,17,19);1H

InChI key

RVFUNJWWXKCWNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

D2 antagonist; 5-HT3 antagonist; antipsychotic; anti-emetic.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Baxter. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R W Pero et al.
Molecular and cellular biochemistry, 193(1-2), 119-125 (1999-05-20)
Our laboratory has concentrated on the possible regulation the benzamides and nicotinamides may have on the processes of DNA repair and apoptosis. Recent reports have suggested that both apoptosis and inflammation are regulated by the transcription factor NF-kappaB. We have
B Y De Winter et al.
Gut, 45(5), 713-718 (1999-10-13)
The effects of different prokinetic agents, the motilide erythromycin and the substituted benzamides metoclopramide and cisapride, were investigated in a rat model of postoperative ileus. These effects were compared with that of granisetron, a 5-hydroxytryptamine (5-HT(3)) receptor antagonist, and a
M K Ladola et al.
Indian journal of pharmaceutical sciences, 76(5), 423-429 (2014-11-27)
In the present investigation, a novel multifunctional co-processed superdisintegrants consisting of crospovidone and Kyron T-314 were fabricated by solvent evaporation method to develop melt-in-mouth tablets of metoclopramide hydrochloride with a view to enhance patient compliance by direct compression method. The
Diogo F Antunes et al.
Molecular and cellular endocrinology, 550, 111649-111649 (2022-04-19)
Dopamine is part of the reward system triggering the social decision-making network in the brain. It has hence great potential importance in the regulation of social behaviour, but its significance in the control of behaviour in highly social animals is
Jonne Rietdijk et al.
BMC biology, 19(1), 156-156 (2021-08-03)
The emergence and continued global spread of the current COVID-19 pandemic has highlighted the need for methods to identify novel or repurposed therapeutic drugs in a fast and effective way. Despite the availability of methods for the discovery of antiviral

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service