Merck
  • Development of liposomal and microemulsion formulations for transdermal delivery of clonazepam: effect of randomly methylated β-cyclodextrin.
Development of liposomal and microemulsion formulations for transdermal delivery of clonazepam: effect of randomly methylated β-cyclodextrin.
International journal of pharmaceutics (2014-09-10)
Paola Mura, Marco Bragagni, Natascia Mennini, Marzia Cirri, Francesca Maestrelli
ABSTRACT
Transdermal administration of clonazepam, a poorly water-soluble benzodiazepine, is an interesting strategy for overcoming the drawbacks of its oral administration. With this aim, two nano-carrier formulations, based on ultra-deformable liposomes and microemulsions, have been developed to favour clonazepam transdermal delivery. Considering the solubilizing power of methyl-βcyclodextrin (Me-βCD) toward clonazepam and its potential positive influence on transdermal drug delivery, the effect of its addition to these formulations was investigated. Artificial lipophilic membranes simulating the skin allowed a rapid evaluation of the drug permeation properties from the systems, compared with those from an aqueous drug suspension, with or without Me-βCD. The best formulations were further characterized by permeation through excised rabbit ear skin. All the formulations increased drug permeability, ranging from 2-fold (liposomes without Me-βCD), up to over 4-fold (microemulsions containing Me-βCD). The different formulations allowed for pointing out different possible permeation enhancing mechanisms of Me-βCD: increase in drug solubility and thermodynamic activity in the vehicle, when added to the drug aqueous suspension; interactions with the vesicle bilayer, in case of liposomal formulations; interactions with the skin membrane lipids, as evidenced in experiments with excised rabbit ear for microemulsions containing Me-βCD, that were then selected for further in vivo studies.
Product Description
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Isopropyl myristate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Triethylamine, purum, ≥99% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Isopropyl myristate, ≥98%
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Octadecylamine, ≥99.0% (GC)
Sigma-Aldrich
Isopropyl myristate, 98%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Octadecylamine, ≥99% (GC)
Sigma-Aldrich
Octadecylamine, technical grade, 90%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Isopropyl myristate, ≥90% (GC)
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Isopropyl myristate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Propylene glycol, European Pharmacopoeia (EP) Reference Standard