Merck
  • The Maillard reaction of bisoprolol fumarate with various reducing carbohydrates.
The Maillard reaction of bisoprolol fumarate with various reducing carbohydrates.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2014-04-22)
Mateusz Szalka, Jacek Lubczak, Dorota Naróg, Marek Laskowski, Krzysztof Kaczmarski
ABSTRACT
HPLC analysis of drug products containing bisoprolol fumarate and lactose revealed the presence of N-formylbisoprolol, which is a final product of the Maillard reaction. Formulations containing secondary amines and reducing carbohydrates are prone to the condensation of amine and carbonyl functional groups and formation of glycosylamines in pharmaceutically relevant conditions. Further rearrangement occurs in the presence of a nucleophile and leads to the formation of 1-deoxy-1-amino-2-ketose also known as the Amadori Rearrangement Product (ARP). The influence of water content, carbohydrate, and lubricant types on the reaction rate was tested. The reaction progress was monitored by HPLC and UV-Vis spectrophotometry. The structures of intermediates were confirmed by the LC/MS(2) analysis. N-formylbisoprolol - the final reaction product - was synthesised and characterised by LC/MS(2), H(1) and C(13) NMR.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Lactose (Monohydrate), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Maltose Monohydrate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Lactose monohydrate, United States Pharmacopeia (USP) Reference Standard
USP
Stearic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Triethylamine, purum, ≥99% (GC)
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
D-Lactose monohydrate, tested according to Ph. Eur.
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
D-Lactose monohydrate, ≥98.0% (HPLC)
Sigma-Aldrich
D-Lactose monohydrate, ACS reagent
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology