Merck
  • O,N,N-pincer ligand effects on oxidatively induced carbon-chlorine coupling reactions at palladium.
O,N,N-pincer ligand effects on oxidatively induced carbon-chlorine coupling reactions at palladium.
Dalton transactions (Cambridge, England : 2003) (2015-02-28)
Luka A Wright, Eric G Hope, Gregory A Solan, Warren B Cross, Kuldip Singh
ABSTRACT
The syntheses of two families of sterically tuneable O,N,N pro-ligands are reported, namely the 2-(phenyl-2'-ol)-6-imine-pyridines, 2-(C6H4-2'-OH),6-(CMe=NAr)C5H3N [Ar = 4-i-PrC6H4 (HL1(a)), 2,6-i-Pr2C6H3 (HL1(b))] and the 2-(phenyl-2'-ol)-6-(amino-prop-2-yl)pyridines, 2-(C6H4-2'-OH),6-(CMe2NHAr)C5H3N [Ar = 4-i-PrC6H4 (HL2(a)), 2,6-i-Pr2C6H3 (HL2(b))], using straightforward synthetic approaches and in reasonable overall yields. Interaction of HL1(a/c) and HL2(a/b) with palladium(II) acetate affords the O,N,N-pincer complexes, [{2-(C6H4-2'-O)-6-(CMe=NAr)C5H3N}Pd(OAc)] (Ar = 4-i-PrC6H4 (1a), 2,6-i-Pr2C6H3 (1b)) and [{2-(C6H4-2'-O)-6-(CMe2NHAr)C5H3N}Pd(OAc)] (Ar = 4-i-PrC6H4 (2a), 2,6-i-Pr2C6H3 (2b)), which can be readily converted to their chloride derivatives, [{2-(C6H4-2'-O)-6-(CMe=NAr)C5H3N}PdCl] (Ar = 4-i-PrC6H4 (3a), 2,6-i-Pr2C6H3 (3b)) and [{2-(C6H4-2'-O)-6-(CMe2NHAr)C5H3N}PdCl] (Ar = 4-i-PrC6H4 (4a), 2,6-i-Pr2C6H3 (4b)), respectively, on reaction with an aqueous sodium chloride solution. Treating each of 3a, 3b, 4a and 4b with two equivalents of di-p-tolyliodonium triflate at 100 °C in a toluene/acetonitrile mixture affords varying amounts of 4-chlorotoluene along with the 4-iodotoluene by-product with the conversions highly dependent on the steric and backbone properties of the pincer complex employed (viz. 4a > 3a > 4b > 3b); notably, the least sterically bulky and most flexible amine-containing 4a reaches 90% conversion to 4-chlorotoluene in 15 h as opposed to 17% for imine-containing 3b. In the case of 3a, the inorganic palladium species recovered from the reaction has been identified as the Pd(II) salt [{2-(C6H4-2'-O)-6-(CMe=N(4-i-PrC6H4)C5H3N}Pd(NCMe)][O3SCF3] (5a), which was independently prepared by the reaction of 3a with silver triflate in acetonitrile. Single crystal X-ray structures are reported for HL1(a), HL2(a), 1a, 1b, 2a, 2b, 3a and 5a.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
4-Isopropylaniline, 99%
Sigma-Aldrich
2,6-Diisopropylaniline, technical grade, 90%
Sigma-Aldrich
2,6-Diisopropylaniline, 97%
Sigma-Aldrich
N-Bromoacetamide, powder
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Supelco
Acetonitrile, analytical standard