Merck
  • Altered plasma lysophosphatidylcholines and amides in non-obese and non-diabetic subjects with borderline-to-moderate hypertriglyceridemia: a case-control study.
Altered plasma lysophosphatidylcholines and amides in non-obese and non-diabetic subjects with borderline-to-moderate hypertriglyceridemia: a case-control study.
PloS one (2015-04-10)
Sae Young Lee, Minjoo Kim, Saem Jung, Sang-Hyun Lee, Jong Ho Lee
ABSTRACT
Hypertriglyceridemia (HTG) is a risk factor for atherosclerotic cardiovascular disease (CVD). We investigated alterations in plasma metabolites associated with borderline-to-moderate HTG (triglycerides (TG) 150-500 mg/dL). Using UPLC-LTQ-Orbitrap mass spectrometry analysis, the metabolomics profiles of 111 non-diabetic and non-obese individuals with borderline-to-moderate HTG were compared with those of 111 age- and sex-matched controls with normotriglyceridemia (NTG, TG <150 mg/dL). When compared to the NTG control group, the HTG group exhibited higher plasma levels of lysophosphatidylcholines (lysoPCs), including C14:0 (q = 0.001) and C16:0 (q = 1.8E-05), and several amides, including N-ethyldodecanamide (q = 2.9E-05), N-propyldodecanamide (q = 3.5E-05), palmitoleamide (q = 2.9E-06), and palmitic amide (q = 0.019). The metabolomic profiles of the HTG group also exhibited lower plasma levels of cis-4-octenedioic acid (q<1.0E-9) and docosanamide (q = 0.002) compared with those of the NTG controls. LysoPC 16:0 and palmitoleamide emerged as the primary metabolites able to discriminate the HTG group from the NTG group in a partial least-squares discriminant analysis and were positively associated with the fasting triglyceride levels. We identified alterations in lysoPCs, amides, and cis-4-octenedioic acid among non-diabetic and non-obese individuals with borderline-to-moderate HTG. These results provide novel insights into the metabolic alterations that occur in the early metabolic stages of HTG. This information may facilitate the design of early interventions to prevent disease progression.
Product Description
Supelco
Sulfadimethoxine, VETRANAL®, analytical standard
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Sulfadimethoxine, 98.0-102.0%
Sulfadimethoxine for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sulfadimethoxine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Terfenadine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Reserpine, crystallized, ≥99.0% (HPLC)
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Reserpine