Skip to Content
Merck
All Photos(1)

Key Documents

PHR1448

Supelco

Sulfadimethoxine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Sulfadimethoxine, 4-Amino-N-(2,6-dimethoxy-4-pyrimidinyl)benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N4O4S
CAS Number:
Molecular Weight:
310.33
Beilstein:
306856
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1626001

API family

sulfadimethoxine

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

COc1cc(NS(=O)(=O)c2ccc(N)cc2)nc(OC)n1

InChI

1S/C12H14N4O4S/c1-19-11-7-10(14-12(15-11)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)

InChI key

ZZORFUFYDOWNEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Sulfadimethoxine is an long acting sulfonamide antibiotic, commonly used against a broad spectrum of bacterial infections in animals. Its mode of action involves the inhibition of folic acid synthesis in prokaryotes. It is also widely used in veterinary medication to treat coccidiosis in cats and dogs.

Application

Sulfadimethoxine may be used as a pharmaceutical reference standard for the quantification of the analyte in veterinary formulations using liquid chromatography technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA2825 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of sulfamethoxypyridazine, sulfamethoxazole, sulfadimethoxine and their associated compounds by liquid chromatography.
Nevado JJ, et al.
Analytica Chimica Acta, 442(2), 241-248 (2001)
Small Animal Clinical Pharmacology (2011)
Minjoo Kim et al.
PloS one, 10(3), e0119519-e0119519 (2015-03-18)
Existing data on the association between being overweight and cardiovascular morbidity and mortality risk in adults are inconsistent. We prospectively and longitudinally investigated the effects of weight on arterial stiffness and plasma metabolites in middle-aged subjects (aged 40-55 years). A
Zhe Meng et al.
Food chemistry, 174, 597-605 (2014-12-23)
A novel UPLC-MS/MS method for quantification and confirmation of eight fluoroquinolones, five sulphonamides (SAs) and four acetyled metabolites in milk is developed. Their main fragmentation pathways were presented. The limits of quantification were in the range of 0.01-0.29 μg kg(-1)
Werner Siegmund et al.
British journal of clinical pharmacology, 79(3), 501-513 (2014-09-30)
The rare association of flupirtine with liver injury is most likely caused by reactive quinone diimines and their oxidative formation may be influenced by the activities of N-acetyltransferases (NAT) that conjugate the less toxic metabolite D13223, and by glucuronosyltransferases (UGT)

Articles

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service