Merck
  • Development of a purity control strategy for pemetrexed disodium and validation of associated analytical methodology.
Development of a purity control strategy for pemetrexed disodium and validation of associated analytical methodology.
Journal of pharmaceutical and biomedical analysis (2014-12-22)
Anne Warner, Irina Piraner, Heather Weimer, Kevin White
ABSTRACT
Stability-indicating reversed phase HPLC methods have been developed and validated for the determination of 13 potential process and degradation impurities in pemetrexed disodium drug substance (DS) and pemetrexed for injection drug product (DP). This paper describes the development of HPLC-UV impurity methods for drug substance and drug product. Relative response factors (RRF) have been determined using HPLC-UV in tandem with CAD or by NMR detection. Conditions for the generation of system suitability solutions are described and assure adequate chromatographic resolution and peak identification without the need for impurity reference standards. The methods were fully validated and demonstrated to have acceptable specificity, linearity, accuracy, repeatability, intermediate precision, detection/quantitation limit, and robustness.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetonitrile-d3, ≥99.8 atom % D, anhydrous
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Acetonitrile-d3, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Acetonitrile-d3, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Acetonitrile-d3, ≥99.8 atom % D
Sigma-Aldrich
Acetonitrile-d3, "100%", 99.96 atom % D
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Acetonitrile-d3, 96-97 atom % D, D2O 15-20 %
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%