Merck
  • Dextrin-colistin conjugates as a model bioresponsive treatment for multidrug resistant bacterial infections.
Dextrin-colistin conjugates as a model bioresponsive treatment for multidrug resistant bacterial infections.
Molecular pharmaceutics (2014-11-02)
Elaine L Ferguson, Ernest Azzopardi, Jessica L Roberts, Timothy R Walsh, David W Thomas
ABSTRACT
Polymer therapeutics offer potential benefits in the treatment of multidrug resistant (MDR) infections; affording targeted delivery of biologically active agents to the site of inflammation, potential decreases in systemic toxicity, and the retention of antimicrobial activity at the target site. As a prototype model, these studies developed and characterized a library of dextrin-colistin conjugates (dextrin molecular weight: 7500-48,000 g/mol) as a means of targeting the delivery of colistin. Optimum colistin release kinetics (following dextrin degradation by physiological concentrations of amylase (100 IU/L)) were observed in conjugates containing low molecular weight (∼7500 g/mol) dextrin with ∼1 mol % succinoylation (∼80% drug release within 48 h, compared to ∼33% from sodium colistin methanesulfonate (CMS, Colomycin)). These conjugates exhibited comparable antimicrobial activity to CMS in conventional MIC assays against a range of Gram-negative pathogens, but with significantly reduced in vitro toxicity toward kidney (IC₅₀ = CMS, 15.4 μg/mL; dextrin-colistin, 63.9 μg/mL) and macrophage (IC₅₀ = CMS, 111.3 μg/mL; dextrin-colistin, 303.9 μg/mL) cells. In vivo dose-escalation studies in rats demonstrated improved pharmacokinetics of the conjugates, with prolonged plasma levels of colistin (t₁/₂ 135-1271 min vs 53 min) and decreased toxicity, compared to colistin sulfate. These studies highlight the potential utility of "nanoantibiotic" polymer therapeutics to aid the safe, effective, and targeted delivery of colistin in the management of MDR infections.
Product Description
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.995% trace metals basis
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Ninhydrin, ACS reagent
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Ninhydrin, suitable for amino acid detection
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture