Merck
  • Synthesis of homopolypeptides by aminolysis mediated by proteases encapsulated in silica nanospheres.
Synthesis of homopolypeptides by aminolysis mediated by proteases encapsulated in silica nanospheres.
Macromolecular bioscience (2014-08-27)
Peter J Baker, Siddharth V Patwardhan, Keiji Numata
ABSTRACT
We report the encapsulation of three different proteases in bioinspired silica. Silica particles were formed under mild reaction conditions using cationic amine-rich ethyleneamines as initiators, which resulted in aggregations of nanoscale spheres. Following encapsulation, the proteases were characterized for their hydrolytic and aminolytic activities. The encapsulation resulted in an increase in the thermal stability of the proteases for both hydrolysis and aminolysis reactions. The enhanced thermal stability of the encapsulated proteases increased the production of poly-L-leucine by aminolysis. Furthermore, the encapsulation of papain resulted in an increase in the production of poly-L-alanine and poly-L-valine at 50 °C.
Product Description
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Chloroethane solution, 2.0 M in tert-butyl methyl ether, anhydrous
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
Pentaethylenehexamine, technical grade
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Supelco
Chloroethane solution, certified reference material, TraceCERT®, 1000 μg/mL in methanol, ampule of 1 mL
Sigma-Aldrich
Tetraethylenepentamine, technical grade
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C
Sigma-Aldrich
Triethylenetetramine, ≥97.0% (T)
Sigma-Aldrich
Triethylenetetramine, technical grade, 60%