Skip to Content
Merck
All Photos(2)

Key Documents

T11509

Sigma-Aldrich

Tetraethylenepentamine

technical grade

Synonym(s):

TEPA, Tetrene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2CH2CH2NHCH2CH2)2NH
CAS Number:
Molecular Weight:
189.30
Beilstein:
506966
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

vapor density

6.53 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥30%

form

viscous liquid

autoignition temp.

610 °F

refractive index

n20/D 1.505 (lit.)

bp

340 °C

mp

−40 °C (lit.)

density

0.998 g/mL at 25 °C (lit.)

SMILES string

NCCNCCNCCNCCN

InChI

1S/C8H23N5/c9-1-3-11-5-7-13-8-6-12-4-2-10/h11-13H,1-10H2

InChI key

FAGUFWYHJQFNRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetraethylenepentamine, also known as Tetrene, is a N5-structure polyamines ligand, often used as a nitrogen precursor to synthesize metal catalysts during oxygen reduction reaction (ORR).

Application

Tetraethylenepentamine (TEPA) can be used as a reagent:      
  • To functionalize magnesium 2,5-dihydroxyterephthalate (Mg-MOF-74) to enhance the CO2 adsorption performance of the material.
  • To modify magnetic chitosan resin to form amine-bearing chitosan for the efficient removal of uranium from an aqueous solution.
  • To synthesize poly(vinyl-chloride)/tetraethylenepentamine (PVC-TEPA) composite material, which is used as an efficient catalyst for the Knoevenagel condensation reaction.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

325.4 °F - closed cup

Flash Point(C)

163 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao Su et al.
ACS applied materials & interfaces, 9(12), 11299-11306 (2017-03-01)
Postsynthetic functionalization of magnesium 2,5-dihydroxyterephthalate (Mg-MOF-74) with tetraethylenepentamine (TEPA) resulted in improved CO2 adsorption performance under dry and humid conditions. XPS, elemental analysis, and neutron powder diffraction studies indicated that TEPA was incorporated throughout the MOF particle, although it coordinated
Khalid Z Elwakeel et al.
Bioresource technology, 160, 107-114 (2014-02-08)
Chitosan was cross-linked using glutaraldehyde in the presence of magnetite. The resin was chemically modified through the reaction with tetraethylenepentamine (TEPA) to produce amine bearing chitosan. The resin showed a higher affinity towards the uptake of UO2(2+) ions from aqueous
Yamin Liu et al.
Environmental science & technology, 45(13), 5710-5716 (2011-06-16)
A novel solid amine sorbent was prepared using KIT-6-type mesoporous silica modified with tetraethylenepentamine (TEPA). Its adsorption behavior toward CO(2) from simulated flue gases is investigated using an adsorption column. The adsorption capacities at temperatures of 303, 313, 333, 343
Takuto Hasegawa et al.
Bioorganic & medicinal chemistry, 17(16), 6015-6019 (2009-07-21)
Two kinds of rhodamine modified beta-cyclodextrins (R-1 and R-2), which are coupled up ethylene diamine (EDA) and tetraethylene pentamine (TEPA) between Rh B and beta-cyclodextrin, respectively, have been synthesized. R-1 and 2 work as a new fluorogenic probe for monitoring
Yu Zhou et al.
Journal of hazardous materials, 190(1-3), 87-93 (2011-03-29)
Novel phenol-capturer was prepared by modifying the as-synthesized mesoporous silica MCM-41 with tetraethylenepentamine (TEPA), not only saving the energy and time for removal of template, but also opening the way to utilize the micelles for adsorption. Once the organic modifier

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service