Skip to Content
Merck
All Photos(4)

Documents

132098

Sigma-Aldrich

Triethylenetetramine

technical grade, 60%

Synonym(s):

DEH 24, EPH 925

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2(NHCH2CH2)2NH2
CAS Number:
Molecular Weight:
146.23
Beilstein:
605448
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39030302
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

vapor density

~5 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

form

liquid

concentration

60%

refractive index

n20/D 1.496 (lit.)

bp

266-267 °C (lit.)

mp

12 °C (lit.)

solubility

alcohol: soluble
water: soluble

density

0.982 g/mL at 25 °C (lit.)

SMILES string

NCCNCCNCCN

InChI

1S/C6H18N4/c7-1-3-9-5-6-10-4-2-8/h9-10H,1-8H2

InChI key

VILCJCGEZXAXTO-UHFFFAOYSA-N

Gene Information

human ... TERT(7015)

Looking for similar products? Visit Product Comparison Guide

General description

Triethylenetetramine is used as a chelating agent and as a curing agent for epoxy resins.

Application


  • Triethylenetetramine in surface passivation: Research on the photooxidation inhibition of pyrite highlights the role of DTC-TETA (dithiocarbamate-triethylenetetramine) in modifying photogenerated carrier migration pathways, indicating its potential in environmental chemistry and surface science (Shu et al., 2024).

  • Poly(urethane-urea) microcapsules crosslinked by triethylenetetramine: An investigation into poly(urethane-urea) microcapsules revealed that triethylenetetramine as a crosslinker can modulate the release of pesticides, providing insights into controlled release systems in agricultural sciences (Wang et al., 2024).

Features and Benefits

Triethylenetetramine is room or low temperature curing agent.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

264.2 °F

Flash Point(C)

129 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvothermal Synthesis of Copper Indium Diselenide in Toluene.
Chang J, et al.
ChemInform, 42(23), 434-438 (2011)
Insights into pore surface modification of mesoporous polymer-silica composites: introduction of reactive amines.
Guillet-Nicolas R, et al.
New. J. Chem., 34(2), 355-366 (2010)
Modified epoxy coatings on mild steel: tribology and surface energy
Brostow W, et al.
European Polymer Journal, 46(11), 2181-2189 (2010)
Dehydration of heavy crude oil emulsions using novel imidazolium-based poly ionic liquids
Ezzat AO, et al.
Journal of Molecular Liquids, 326, 115284-115284 (2021)
Synthesis of epoxy resins derivatives of naphthalene-2, 7-diol and their cross-linked products
Fila K, et al.
Journal of Thermal Analysis and Calorimetry, 138, 4349-4358 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service