Merck
  • Mianserin, an antidepressant kills Leishmania donovani by depleting ergosterol levels.
Mianserin, an antidepressant kills Leishmania donovani by depleting ergosterol levels.
Experimental parasitology (2014-06-21)
Neeradi Dinesh, Preet Kamal Kaur, Kayala Kambagiri Swamy, Sushma Singh
ABSTRACT
In the present study, we have investigated the antileishmanial potential of mianserin, an antidepressant. Mianserin was found to inhibit both the promastigote and amastigote forms of the parasite in a dose dependant manner. The IC50 values for promastigotes and amastigotes were 21 μM and 46 μM respectively. Interestingly, mianserin failed to inhibit THP-1 differentiated macrophages up to 100 μM concentration thus, exhibiting parasite selectivity. When mianserin was incubated with recombinant Leishmania donovani 3-hydroxy-3-methylglutaryl coenzyme A reductase (HMGR) enzyme, it exhibited an IC50 value of 19.8 μM. Inhibition kinetics revealed competitive mode of enzyme inhibition as the Km increased with no change in Vmax. Further structural investigation of enzyme-inhibitor interaction revealed quenching of HMGR tryptophan intrinsic fluorescence with a K(sv) value of 3.025±0.37 M(-1) and an apparent binding constant of 0.0954 mM. We further estimated ergosterol levels which is a major component of Leishmania cell membrane. It is synthesized by HMGR enzyme, the first rate limiting enzyme of the sterol biosynthetic pathway. Analysis of ergosterol levels by HPLC revealed ∼2.5-fold depletion in mianserin treated promastigotes with respect to untreated parasites. This data was further validated by exogenous supplementation of mianserin treated cells with ergosterol and cholesterol. Reversal of growth inhibition was observed only upon ergosterol addition though it was refractory to cholesterol supplementation. Overall, our results demonstrate the possibility of repositioning of an antidepressant for the treatment of Visceral Leishmaniasis.
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
SAFC
HEPES
Mianserin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
HEPES
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Ergosterol, ≥75%
Sigma-Aldrich
Mianserin hydrochloride
Mianserin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Ergosterol, European Pharmacopoeia (EP) Reference Standard
Supelco
Ergosterol, Pharmaceutical Secondary Standard; Certified Reference Material