Skip to Content
Merck
All Photos(1)

Documents

PHR1428

Supelco

HEPES

certified reference material,pharmaceutical secondary standard

Synonym(s):

HEPES, 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O4S
CAS Number:
Molecular Weight:
238.30
Beilstein:
883043
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

product name

HEPES, Pharmaceutical Secondary Standard; Certified Reference Material

grade

certified reference material
pharmaceutical secondary standard

Quality Level

API family

hepes

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

pH

5.0-6.5 (25 °C, 238 g/L)

useful pH range

6.8-8.2

pKa (25 °C)

7.5

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

OCCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)

InChI key

JKMHFZQWWAIEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

HEPES has been described as one of the best all-purpose buffers available for biological research. At biological pH, the molecule is zwitterionic and is effective as a buffer at pH 6.8 to 8.2.


Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
  • HEPES has been used in a wide variety of applications, including tissue culture.
  • It is used to buffer cell culture media in air.
  • Finds its usage in invitro experiments on Mg.
  • Recognized as one of Dr.Good′s recommended buffers, used in some cell culture media as a buffering agent and also in various biochemical reactions.
  • Recently, in the production of radiopharmaceuticals, it is the buffer of choice for scientific labeling.

HEPES may also be used as:
  • Component of homogenization buffer to homogenize adipose tissue for liquid chromatography/mass spectrometry and in HN buffer for dissolving viral stock titer after dot blot hybridization.
  • Pharmaceutical reference standard for the determination of the analyte in bulk drug and pharmaceutical formulations by high performance liquid chromatography.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3253 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of HEPES in 68 Ga-labeled peptide solutions
Sasson R, et al.
J. Radioanal. Nucl. Chem., 283(3), 753- 756 (2010)
Simple and reliable chromatographic technique for the determination of HEPES in Ga-68-labeled pharmaceuticals
Maus S, et al.
Journal of Nuclear Medicine, 56(3), 1168- 1168 (2015)
Development and evaluation of a rapid analysis for HEPES determination in 68 Ga-radiotracers
Pfaff S, et al.
EJNMMI Research, 8(1), 95-95 (2018)
FGF21 Prevents Angiotensin II-Induced Hypertension and Vascular Dysfunction by Activation of ACE2/Angiotensin-(1?7) Axis in Mice.
Pan X, et al.
Cell Metabolism (2018)
Hydrogen ion buffers for biological research.
Good NE, et al.
Biochemistry, 5(2), 467-477 (1966)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service