Skip to Content
Merck
All Photos(4)

Key Documents

M2525

Sigma-Aldrich

Mianserin hydrochloride

Synonym(s):

1,2,3,4,10,14b-Hexahydro-2-methyl-dibenzo[c,f]pyrazino[1,2-a]azepine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20N2 · HCl
CAS Number:
Molecular Weight:
300.83
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

solubility

H2O: 3.4 mg/mL
ethanol: 5.6 mg/mL

originator

Organon

storage temp.

2-8°C

SMILES string

Cl.CN1CCN2C(C1)c3ccccc3Cc4ccccc24

InChI

1S/C18H20N2.ClH/c1-19-10-11-20-17-9-5-3-7-15(17)12-14-6-2-4-8-16(14)18(20)13-19;/h2-9,18H,10-13H2,1H3;1H

InChI key

YNPFMWCWRVTGKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Mianserin hydrochloride has been used:
  • as a reversible antagonist for serotonergic -protein coupled receptor (GPCR) - G-protein protein-coupled receptor (S7.1R)
  • as an antidepressant in hippocampal astrocytes to test its effect on brain-derived neurotrophic factor (BDNF) mRNA expression
  • as a 5-hydroxytryptamine (5-HT) receptor antagonist to study its effect on serotonin modulation

Biochem/physiol Actions

Antidepressant; antagonist/inverse agonist at 5-HT2 serotonin receptors; also blocks the H1 histamine receptor and the α2 adrenoceptor.
Mianserin is a tetracyclic compound with sedative property. It lacks anticholinergic activity and is not effective on serotoninergic mechanisms. Mianserin is eliminated post biotransformation as hydroxylation, N-oxidation and N-demethylation products.

Features and Benefits

This compound was developed by Organon. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Solutions may be stored for several days at 4°C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T de Boer
International clinical psychopharmacology, 10 Suppl 4, 19-23 (1995-12-01)
Mirtazapine is a new antidepressant with a unique mode of action: it preferentially blocks the noradrenergic alpha2-auto- and heteroreceptors held responsible for controlling noradrenaline and serotonin release. In addition, mirtazapine has a low affinity for serotonin (5-HT)1A receptors but potently
Modulatory effects of the serotonergic and histaminergic systems on reaction to light in the crustacean Gammarus pulex
Perrot-Minnot MJ, et al.
Neuropharmacology, 75, 31-37 (2013)
I M Leitch et al.
The Journal of pharmacy and pharmacology, 44(4), 315-320 (1992-04-01)
Some in-vitro pharmacological effects of a novel analogue of mianserin, 2-carboxamidino-1,2,3,4,10,14b-hexahydrodibenzo (c,f) pyrazino (1,2-alpha) azepine hydrochloride (FCC5) have been studied. FCC5 was a non-competitive antagonist of both histamine-induced contractions of the guinea-pig ileum and 5-HT-induced contractions of rat fundal strips
P A Lavoie et al.
European journal of pharmacology, 369(3), 387-390 (1999-05-04)
In order to test potential links with other stereospecific neurobiological effects of mianserin, the present study explored the stereospecificity for inhibition of depolarization-induced Ca2+ uptake by mianserin. Synaptosomes from rat hippocampus were incubated with 45Ca2+ in either resting or depolarizing
Mianserin: a review of its pharmacological properties and therapeutic efficacy in depressive illness
Brogden RN, et al.
Drugs, 16(4), 273-301 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service