Merck
  • Antiallergic and antiasthmatic effects of a novel enhydrazinone ester (CEE-1): inhibition of activation of both mast cells and eosinophils.
Antiallergic and antiasthmatic effects of a novel enhydrazinone ester (CEE-1): inhibition of activation of both mast cells and eosinophils.
The Journal of pharmacology and experimental therapeutics (2014-06-12)
Charles I Ezeamuzie, Ahmed Z El-Hashim, Waleed M Renno, Ivan O Edafiogho
ABSTRACT
Activation of mast cells and eosinophils is a fundamental process in the pathophysiology of allergic diseases. We have previously reported that the novel enhydrazinone ester CEE-1 (ethyl 4-phenylhydrazinocyclohex-3-en-2-oxo-6-phenyl-1-oate) possesses potent anti-inflammatory activity. We have now tested whether the compound also possesses antiallergic and antiasthmatic effects in vitro and in vivo. The compound significantly inhibited degranulation and leukotriene C4 (LTC4) release from activated human eosinophils, as well as IgE-dependent degranulation and LTC4 release from passively sensitized rat basophilic leukemia cells and bone marrow-derived mouse mast cells. In human eosinophils, the drug was more potent in inhibiting degranulation than LTC4 release {IC50 = 0.4 μM [confidence interval (CI): 0.1-0.9] versus 3.8 μM (CI: 0.9-8.3)}, whereas in mast cells the reverse was essentially the case. The drug did not affect stimulus-induced calcium transients in eosinophils but significantly inhibited early phosphorylation of extracellular signal-regulated kinases 1/2 and p38-mitogen-activated protein kinases (MAPK). In vivo, topical application of 4.5-15 mg/kg of the compound significantly inhibited allergen-induced passive cutaneous anaphylaxis in mice. Similarly, in the mouse asthma model, the intranasal administration of 6.5-12.5 mg/kg of the compound significantly inhibited bronchial inflammation and eosinophil accumulation in bronchial lavage fluid, as well as abolishing airway hyper-responsiveness to methacholine. These results show that CEE-1 inhibits the activation of both mast cells and eosinophils in vitro, probably by blocking MAPK-activation pathways, and that these effects are translated into antiallergic and antiasthmatic effects in vivo. The compound, therefore, has potential application in the treatment of asthma and other allergic diseases.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Acetone, analytical standard
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, for molecular biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder