Skip to Content
Merck
All Photos(1)

Key Documents

420883

Sigma-Aldrich

LUDOX® CL colloidal silica

30 wt. % suspension in H2O

Synonym(s):

Silica preparation

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352309
PubChem Substance ID:
NACRES:
NA.22

description

colloidal

form

suspension

composition

SiO2 +Al2O3

concentration

30 wt. % suspension in H2O

surface area

~230 m2/g

pH

4.5

density

1.23 g/mL at 25 °C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Alumina coated chloride stabilizing counterion

Legal Information

LUDOX is a registered trademark of W.R. Grace & Co.-Conn.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David M Brown et al.
Toxicology letters, 224(1), 147-156 (2014-01-28)
We investigated the effects of silica particles and nanoparticles (NPs) (50 nm and 200 nm) with a neutral and positively charged surface when dispersed in saline, bovine serum albumin (BSA) or lung lining fluid (LLF) 24 h post instillation into
Fabrice Gritti et al.
Journal of chromatography. A, 1342, 24-29 (2014-04-17)
The advantages of using columns packed with stationary phases having a composition gradient so that retention factors increase toward the outlet and eluting them with an isocratic mobile phase may provide increased peak resolution in liquid chromatography. This approach is
Richard Hayes et al.
Journal of chromatography. A, 1357, 36-52 (2014-05-27)
The challenges in HPLC are fast and efficient separation for a wide range of samples. Fast separation often results in very high operating pressure, which places a huge burden on HPLC instrumentation. In recent years, core-shell silica microspheres (with a
Alexandra I Lupulescu et al.
Science (New York, N.Y.), 344(6185), 729-732 (2014-05-17)
The growth mechanism of silicalite-1 (MFI zeolite) is juxtaposed between classical models that postulate silica molecules as primary growth units and nonclassical pathways based on the aggregation of metastable silica nanoparticle precursors. Although experimental evidence gathered over the past two
Xiaoxiao He et al.
Journal of biomedical nanotechnology, 10(7), 1359-1368 (2014-05-09)
This paper describes a sensitive and specific determination strategy for Staphylococcus aureus (S. aureus) detection using aptamer recognition and fluorescent silica nanoparticles (FSiNPs) label based dual-color flow cytometry assay (Aptamer/FSiNPs-DCFCM). In the protocol, an aptamer, having high affinity to S.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service