Merck
  • Discovery of cellobionic acid phosphorylase in cellulolytic bacteria and fungi.
Discovery of cellobionic acid phosphorylase in cellulolytic bacteria and fungi.
FEBS letters (2013-09-24)
Takanori Nihira, Yuka Saito, Mamoru Nishimoto, Motomitsu Kitaoka, Kiyohiko Igarashi, Ken'ichi Ohtsubo, Hiroyuki Nakai
ABSTRACT
A novel phosphorylase was characterized as new member of glycoside hydrolase family 94 from the cellulolytic bacterium Xanthomonas campestris and the fungus Neurospora crassa. The enzyme catalyzed reversible phosphorolysis of cellobionic acid. We propose 4-O-β-D-glucopyranosyl-D-gluconic acid: phosphate α-D-glucosyltransferase as the systematic name and cellobionic acid phosphorylase as the short names for the novel enzyme. Several cellulolytic fungi of the phylum Ascomycota also possess homologous proteins. We, therefore, suggest that the enzyme plays a crucial role in cellulose degradation where cellobionic acid as oxidized cellulolytic product is converted into α-D-glucose 1-phosphate and D-gluconic acid to enter glycolysis and the pentose phosphate pathway, respectively.
Sigma-Aldrich
D-Gluconic acid solution, 49-53 wt. % in H2O
Sigma-Aldrich
D-Gluconic acid sodium salt, ≥99% (HPLC)
Supelco
D-Gluconic acid sodium salt, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium gluconate, meets USP testing specifications
Sigma-Aldrich
Iron(II) D-gluconate dihydrate, 98%
Sigma-Aldrich
Magnesium D-gluconate hydrate, ≥98% (HPLC)
Sigma-Aldrich
Magnesium D-gluconate hydrate, 98.0-102% anhydrous basis, meets USP testing specifications
Sigma-Aldrich
Potassium gluconate, 97.0-103.0% dry basis, meets USP testing specifications
Supelco
Potassium gluconate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium D-gluconate, ≥99% (HPLC)