Skip to Content
Merck
All Photos(3)

Key Documents

G9130

Sigma-Aldrich

Magnesium D-gluconate hydrate

≥98% (HPLC)

Synonym(s):

D-Gluconic acid hemimagnesium salt, D-Gluconic acid magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22MgO14 · xH2O
CAS Number:
Molecular Weight:
414.60 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

technique(s)

HPLC: suitable

SMILES string

O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O[Mg]OC(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/2C6H12O7.Mg.H2O/c2*7-1-2(8)3(9)4(10)5(11)6(12)13;;/h2*2-5,7-11H,1H2,(H,12,13);;1H2/q;;+2;/p-2/t2*2-,3-,4+,5-;;/m11../s1

InChI key

IAKLPCRFBAZVRW-XRDLMGPZSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Gluconic acid occurs from the oxidation of glucose and is naturally in fruit, honey and wine. When used as a food additive it helps regulate acidity. It is a strong chelating agent, chelating the anions of calcium, iron, aluminum, copper, and other heavy metals.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suvarna Mini Vijayan et al.
Toxicology in vitro : an international journal published in association with BIBRA, 71, 105055-105055 (2020-11-24)
The fluoride ions of the industrially largely irreplaceable, locally corrosive hydrofluoric acid (HF) can scavenge cations in biological tissues, which explains their high toxic potential, and also leads to local acidification through proton release. The influence of three complexing agents
Jeffrey E Nowak et al.
Pediatric critical care medicine : a journal of the Society of Critical Care Medicine and the World Federation of Pediatric Intensive and Critical Care Societies, 13(5), e323-e329 (2012-07-05)
We previously demonstrated that altered zinc homeostasis is an important feature of pediatric sepsis, thus raising the possibility of zinc supplementation as a therapeutic strategy in sepsis. Herein, we tested the hypothesis that prophylactic zinc supplementation would be beneficial in
Angel A Yanagihara et al.
PloS one, 7(12), e51368-e51368 (2012-12-20)
Chironex fleckeri (Australian box jellyfish) stings can cause acute cardiovascular collapse and death. We developed methods to recover venom with high specific activity, and evaluated the effects of both total venom and constituent porins at doses equivalent to lethal envenomation.
Maria Meyer et al.
Applied microbiology and biotechnology, 97(8), 3457-3466 (2012-07-14)
The acetic acid bacterium Gluconobacter oxydans incompletely oxidizes carbon sources as a natural part of its metabolism, and this feature has been exploited for many biotechnological applications. The most important enzymes used to harness the biocatalytic oxidative capacity of G.
Anja Troge et al.
International journal of medical microbiology : IJMM, 302(7-8), 304-314 (2012-11-08)
The flagellum of the probiotic Escherichia coli strain Nissle 1917 (EcN) is not just responsible for motility, but also for EcN's ability to induce the production of human β-defensin 2. Here, we report a third function of this EcN organell.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service