Skip to Content
Merck
All Photos(4)

Key Documents

P1847

Sigma-Aldrich

Potassium gluconate

97.0-103.0% dry basis, meets USP testing specifications

Synonym(s):

Potassium D-gluconate, 2,3,4,5,6-Pentahydroxycaproic acid potassium salt, D-Gluconic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11KO7
CAS Number:
Molecular Weight:
234.25
Beilstein:
3919448
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

Agency

USP/NF
meets USP testing specifications

Assay

97.0-103.0% dry basis

form

powder

impurities

≥1.0 % (v/v) reducing substances

color

white

mp

183 °C (dec.) (lit.)

solubility

water: soluble

application(s)

pharmaceutical (small molecule)

SMILES string

[K+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

InChI

1S/C6H12O7.K/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h2-5,7-11H,1H2,(H,12,13);/q;+1/p-1/t2-,3-,4+,5-;/m1./s1

InChI key

HLCFGWHYROZGBI-JJKGCWMISA-M

Looking for similar products? Visit Product Comparison Guide

Application

Potassium gluconate has been used as:
  • a component in intracellular solution (1x) for electrophysiology recordings in cultured neurons
  • as a component of solution for electroporation of neurons
  • in the preparation of intracellular pipette solution

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Assembly and operation of the autopatcher for automated intracellular neural recording in vivo
Kodandaramaiah SB, et al.
Nature Protocols, 11(4), 634-634 (2016)
In vivo functional mapping of a cortical column at single-neuron resolution
Tischbirek CH, et al.
Cell Reports, 27(5), 1319-1326 (2019)
Optogenetic stimulation and recording of primary cultured neurons with spatiotemporal control
Barral J and Reyes AD
Bio-protocol, 7(12) (2017)
Carsten H Tischbirek et al.
Cell reports, 27(5), 1319-1326 (2019-05-03)
The cerebral cortex is organized in vertical columns that contain neurons with similar functions. The cellular micro-architecture of such columns is an essential determinant of brain dynamics and cortical information processing. However, a detailed understanding of columns is incomplete, even
Esther Krook-Magnuson et al.
The Journal of physiology, 593(10), 2379-2388 (2015-03-11)
The dentate gyrus is a region subject to intense study in epilepsy because of its posited role as a 'gate', acting to inhibit overexcitation in the hippocampal circuitry through its unique synaptic, cellular and network properties that result in relatively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service