Merck
  • Effects of electron beam irradiation on the migration of antioxidants and their degradation products from commercial polypropylene into food simulating liquids.
Effects of electron beam irradiation on the migration of antioxidants and their degradation products from commercial polypropylene into food simulating liquids.
Food additives and contaminants (1993-07-01)
F Bourges, G Bureau, B Pascat
ABSTRACT
The influence of electron beam irradiation on migrational behaviour of additives present in food packaging material was studied. The migration experiments were carried out on irradiated and non-irradiated polypropylene pouches containing aqueous food simulating liquids (FSL) for 10 days at 40 degrees C. The controls were irradiated and non-irradiated pouches without FSL contact. After the contact period, the polypropylene and the FSL were analysed. A comparison between the results obtained by the two analyses showed the migration of three products of antioxidant degradation from the polypropylene into the FSL, and a partial decomposition of these migrants in the FSL.
Product Description
Plastic additive 12, European Pharmacopoeia (EP) Reference Standard
Plastic additive 09, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tris(2,4-di-tert-butylphenyl) phosphite
Sigma-Aldrich
Pentaerythritol tetrakis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate), 98%