Skip to Content
Merck
All Photos(1)

Key Documents

P2155012

Plastic additive 12

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Tris(2,4-di-tert-butylphenyl) phosphite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[[(CH3)3C]2C6H3O]3P
CAS Number:
Molecular Weight:
646.92
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

mp

181-184 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)(C)c1ccc(OP(Oc2ccc(cc2C(C)(C)C)C(C)(C)C)Oc3ccc(cc3C(C)(C)C)C(C)(C)C)c(c1)C(C)(C)C

InChI

1S/C42H63O3P/c1-37(2,3)28-19-22-34(31(25-28)40(10,11)12)43-46(44-35-23-20-29(38(4,5)6)26-32(35)41(13,14)15)45-36-24-21-30(39(7,8)9)27-33(36)42(16,17)18/h19-27H,1-18H3

InChI key

JKIJEFPNVSHHEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Plastic additive 12 EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ebru Oral et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 32(6), 757-761 (2014-02-19)
Antioxidant stabilization of radiation cross-linked ultrahigh molecular weight polyethylene (UHMWPE) has been introduced to improve the oxidative stability of total joint implant bearing surfaces. Blending of an antioxidant with UHMWPE resin powder followed by consolidation and radiation cross-linking has been
Taoufiq Mouhib et al.
Journal of the American Society for Mass Spectrometry, 21(12), 2005-2010 (2010-09-25)
The enhancement of the static secondary ion mass spectrometry (SIMS) signals resulting from the injection, closely to the sample surface, of H(2)O vapor at relatively high-pressure, was investigated for a set of organic materials. While the ion signals are generally
F Bourges et al.
Food additives and contaminants, 10(4), 443-452 (1993-07-01)
The influence of electron beam irradiation on migrational behaviour of additives present in food packaging material was studied. The migration experiments were carried out on irradiated and non-irradiated polypropylene pouches containing aqueous food simulating liquids (FSL) for 10 days at
K Fischer et al.
Chemosphere, 39(4), 611-625 (1999-07-28)
In order to evaluate the environmental risk potential of the polymer-additives octadecyl-3-(3.5-di-t-butyl-4-hydroxyphenyl)propionate (additive 1) and tri-(2.4-di-t-butylphenyl)phosphite (additive 2) we have studied the degradation of the 14C-labelled single components and their release from a polypropylene matrix in abiotic and biotic test
B Marcato et al.
International journal of pharmaceutics, 257(1-2), 217-225 (2003-04-25)
The migration of the antioxidant additives pentaerythrityl tetrakis(3,5-di-tert-butyl-4-hydroxyphenyl)propionate (Irganox 1010) and tris(2,4-di-tert-butylphenyl)phosphite (Irgafos 168) from polyolefinic packaging into oily vehicles was investigated. The polyolefins included in the study were from the following classes: isotactic polypropylene homopolymer (PP), ethylene-co-propylene random copolymer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service