Skip to Content
Merck
All Photos(1)

Key Documents

P2155009

Plastic additive 09

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Pentaerythritol tetrakis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate), Pentaerythritol tetrakis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HOC6H2[C(CH3)3]2CH2CH2CO2CH2]4C
CAS Number:
Molecular Weight:
1177.63
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

mp

115-118 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)(C)c1cc(CCC(=O)OCC(COC(=O)CCc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)(COC(=O)CCc3cc(c(O)c(c3)C(C)(C)C)C(C)(C)C)COC(=O)CCc4cc(c(O)c(c4)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C

InChI

1S/C73H108O12/c1-65(2,3)49-33-45(34-50(61(49)78)66(4,5)6)25-29-57(74)82-41-73(42-83-58(75)30-26-46-35-51(67(7,8)9)62(79)52(36-46)68(10,11)12,43-84-59(76)31-27-47-37-53(69(13,14)15)63(80)54(38-47)70(16,17)18)44-85-60(77)32-28-48-39-55(71(19,20)21)64(81)56(40-48)72(22,23)24/h33-40,78-81H,25-32,41-44H2,1-24H3

InChI key

BGYHLZZASRKEJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Plastic additive 09 EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T D Lickly et al.
Food additives and contaminants, 7(6), 805-814 (1990-11-01)
Alternatives to highly-volatile ethanol or analytically complex cooking oil were examined as potential fatty-food simulants which would undergo high-temperature exposures to food-packaging polymers in food-packaging evaluation studies. The alternatives consisted of alcohols containing four to eight carbons. As test cases
A Delcorte et al.
Analytical chemistry, 75(24), 6875-6885 (2003-12-13)
The metallization procedure, proposed recently for signal improvement in organic secondary ion mass spectrometry (SIMS) (Delcorte, A.; Médard, N.; Bertrand, P. Anal.Chem. 2002, 74, 4955)., has been thoroughly tested for a set of kilodalton molecules bearing various functional groups: Irganox
[A method for the quantitative determination of phenosan-23].
G A Babenko et al.
Gigiena truda i professional'nye zabolevaniia, (11)(11), 58-58 (1986-11-01)
J L S Lee et al.
Analytical chemistry, 82(1), 98-105 (2009-12-05)
Cluster ion beams have revolutionized the analysis of organic surfaces in time-of-flight secondary ion mass spectrometry and opened up new capabilities for organic depth profiling. Much effort has been devoted to understanding the capabilities and improving the performance of SF(5)(+)
A D Schwope et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 25(4), 327-330 (1987-04-01)
In a series of experiments on the migration of the antioxidant Irganox 1010 from ethylene-vinyl acetate (EVA) films into food-simulating liquids and foods, the antioxidant was found to migrate rapidly from EVA film into n-heptane, 100% ethanol and corn oil.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service