Skip to Content
Merck
All Photos(2)

Key Documents

326542

Sigma-Aldrich

Gold

beads, 1-6 mm, 99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Au
CAS Number:
Molecular Weight:
196.97
EC Number:
MDL number:
UNSPSC Code:
12141717
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

beads

resistivity

2.05 μΩ-cm, 0°C

particle size

1-6 mm

bp

2808 °C (lit.)

mp

1063 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

General description

Goldis a highly valued metal in catalysis and material science due to its uniqueproperties such as high chemical stability, excellent electrical and thermalconductivity, malleability, and ductility. In catalysis, gold nanoparticles areparticularly effective as catalysts for various chemical reactions, includingoxidation and hydrogenation processes due to their high surface area andexcellent catalytic activity. In material science, gold is often employed inthe fabrication of electronic components, sensors, and biomedical devices dueto its excellent conductivity, corrosion resistance, and biocompatibility.

Application

  • Design and performance analysis of a highly sensitive photonic crystal fiber based plasmonic sensor: This study highlights the development of a gold-based plasmonic sensor for applications in material science, particularly for environmental monitoring (Walid et al., 2024).
  • A theoretical study of the electronic structures of metal-semiconductor-metal junction and their electrical properties: Discusses the electronic properties of gold in semiconductor devices, valuable for academic research in material sciences (Almehanyah, 2024).
  • Highly stable and efficient gold nanoparticles films facilitated by thiourea for catalytic reduction of nitrophenol in water: Investigates gold nanoparticles for environmental remediation, an application of interest in material science for its efficiency in catalytic processes (Xia et al., 2024).

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Young Joo Choi et al.
Journal of nanoscience and nanotechnology, 13(6), 4437-4445 (2013-07-19)
Gold nanorods (Au NRs) that absorb near-infrared (NIR) light have great potential in the field of nanomedicine. Photothermal therapy (PTT), a very attractive cancer therapy in nanomedicine, combines nanomaterials and light. The aim of this study was to elucidate the
Zhengxia Liu et al.
Journal of biomedical nanotechnology, 9(6), 1017-1028 (2013-07-19)
Ischemic stroke is a leading cause of death and disability, and the treatment options are limited. Recent studies demonstrated that the promising applications of gold nanoparticles (Au-NPs) as intrinsic therapeutics; however, little is known about the effect of Au-NPs on
Fengxian Zheng et al.
Journal of nanoscience and nanotechnology, 13(6), 3990-3998 (2013-07-19)
Gold nanoparticles (AuNPs) are of biomedical importance, such as delivery vectors. Therefore, we used all-atom molecular dynamic simulations to study the interaction of AuNPs with cell membrane (DMPC bilayer). We observed that the AuNPs adhered spontaneously on the surface of
Matthieu B Miller et al.
Journal of the Air & Waste Management Association (1995), 63(6), 681-693 (2013-07-19)
Industrial gold mining is a significant source of mercury (Hg) emission to the atmosphere. To investigate ways to reduce these emissions, reclamation and dust and mercury control methods used at open pit gold mining operations in Nevada were studied in
Bibek Jyoti Borah et al.
Journal of nanoscience and nanotechnology, 13(7), 5080-5087 (2013-08-02)
Multifunctional phosphine based ligands, 1,1,1-tris(diphenylphosphinomethyl)ethane [CH3C(CH2 PPh2)3][P3] and 1,1,1-tris(diphenylphosphinomethyl)ethane trisulphide [CH3C(CH2P(S)Ph2)3][P3S3] have been introduced to stabilize Au(o)-nanoparticles having small core diameter and narrow size distribution. The Au(o)-nanoparticles were synthesized by the reduction of HAuCl4 precursor with NaBH4 in the presence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service