Merck
  • Fabrication phosphomolybdic acid-reduced graphene oxide nanocomposite by UV photo-reduction and its electrochemical properties.
Fabrication phosphomolybdic acid-reduced graphene oxide nanocomposite by UV photo-reduction and its electrochemical properties.
Physical chemistry chemical physics : PCCP (2013-03-12)
Jie Chen, Suling Liu, Wei Feng, Guoquan Zhang, Fenglin Yang
ABSTRACT
Phosphomolybdic acid-reduced graphene oxide (PMoA-rGO) nanocomposites are fabricated by a photochemical reduction method. They are characterized by ultraviolet-visible spectra, scanning electron microscope images, Fourier transform infrared spectroscopy, Raman spectra and X-ray photoelectron spectroscopy in order to confirm that oxygen-containing functional groups on GO are replaced by PMoA and that GO is reduced to rGO in the photo-reduction process. The electrochemical properties of PMoA-rGO are investigated by cyclic voltammetry, which shows that the PMoA-rGO modified glassy carbon electrode has high electrocatalytic activity in acid solution via a fast, surface-controlled electron transfer process. The results indicate that the use of rGO not only increases the electroactive surface area, but also facilitates electron transfer due to its high electric conductivity.
Sigma-Aldrich
Sodium phosphomolybdate hydrate, technical
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Phosphomolybdic acid solution, 20 wt. % in ethanol
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Phosphomolybdic Acid Solution
Sigma-Aldrich
Phosphomolybdic acid solution, ready-to-use spray and plunge reagent, for chromatography
Sigma-Aldrich
Phosphomolybdic acid solution, spray reagent, 10% in ethanol
Sigma-Aldrich
Molybdenum, wire, diam. 1.0 mm, 99.95% trace metals basis