Skip to Content
Merck
All Photos(2)

Documents

510092

Sigma-Aldrich

Molybdenum

powder, 1-5 μm, ≥99.9% trace metals basis

Synonym(s):

Molybdenum element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mo
CAS Number:
Molecular Weight:
95.94
EC Number:
MDL number:
UNSPSC Code:
12141727
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.9% trace metals basis

form

powder

resistivity

5.0 μΩ-cm, 20°C

particle size

1-5 μm

bp

4612 °C (lit.)

mp

2617 °C (lit.)

density

10.3 g/mL at 25 °C (lit.)

SMILES string

[Mo]

InChI

1S/Mo

InChI key

ZOKXTWBITQBERF-UHFFFAOYSA-N

Application



  • Stress mitigation and cognitive function: A study indicates that venlafaxine, combined with synbiotic supplementation, can ameliorate stress-induced cognitive impairments and improve intestinal barrier integrity, highlighting its therapeutic potential beyond conventional depression treatment (Wongchitrat et al., 2024).


  • Drug degradation pathways: Research explores the degradation of venlafaxine through photoinduced and electrochemically induced methods, providing insights into environmental pathways and degradation kinetics critical for pharmaceutical waste management (Voigt et al., 2024).



Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuclear nonproliferation. Iranian 'medical' reactor stokes search for alternatives.
Richard Stone
Science (New York, N.Y.), 340(6139), 1392-1392 (2013-06-22)
Elena Vigonsky et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(14), 5440-5445 (2013-03-21)
In all kingdoms of life, ATP Binding Cassette (ABC) transporters participate in many physiological and pathological processes. Despite the diversity of their functions, they have been considered to operate by a largely conserved mechanism. One deviant is the vitamin B12
Jie Chen et al.
Physical chemistry chemical physics : PCCP, 15(15), 5664-5669 (2013-03-12)
Phosphomolybdic acid-reduced graphene oxide (PMoA-rGO) nanocomposites are fabricated by a photochemical reduction method. They are characterized by ultraviolet-visible spectra, scanning electron microscope images, Fourier transform infrared spectroscopy, Raman spectra and X-ray photoelectron spectroscopy in order to confirm that oxygen-containing functional
Alexander O Terent'ev et al.
Organic & biomolecular chemistry, 11(16), 2613-2623 (2013-03-01)
Phosphomolybdic acid (PMA) and phosphotungstic acid (PTA) efficiently catalyze the addition of H2O2 to β-diketones to form bridged 1,2,4,5-tetraoxanes. These reactions are not accompanied by the formation of monocyclic peroxides containing hydroxy and hydroperoxide groups or polymeric peroxides. The use
Mădălina Vieriu et al.
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi, 116(4), 1218-1222 (2013-05-25)
Ramipril is a drug of the angiotensin converting enzyme inhibitor class. A new molecular absorbance spectrometric method was developed for the assay of ramipril using molybdophosphoric acid in acidic medium. The reaction product showed a maximum absorbance at 361 nm.

Articles

Permanent magnets are an essential technology for energy conversion. Motors and generators are used to convert energy between electrical and mechanical forms.

Biomedical implants are essentially foreign substances within the human body that must survive many years’ exposure to demanding mechanical and physiological conditions. Despite these challenges, metal implants have been widely used to substitute for or rebuild hard tissues such as bones and teeth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service