Merck
QSAR-based solubility model for drug-like compounds.
Bioorganic & medicinal chemistry (2010-09-03)
Rafael Gozalbes, Antonio Pineda-Lucena
ABSTRACT
Solubility plays a very important role in the selection of compounds for drug screening. In this context, a QSAR model was developed for predicting water solubility of drug-like compounds. First, a set of relevant parameters for establishing a drug-like chemical space was defined. The comparison of chemical structures from the FDAMDD and PHYSPROP databases allowed the selection of properties that were more efficient in discriminating drug-like compounds from other chemicals. These filters were later on applied to the PHYSPROP database and 1174 chemicals fulfilling these criteria and with experimental solubility information available at 25°C were retained. Several QSAR solubility models were developed from this set of compounds, and the best one was selected based on the accuracy of correct classifications obtained for randomly chosen training and validation subsets. Further validation of the model was performed with a set of 102 drugs for which experimental solubility data have been recently reported. A good agreement between the predictions and the experimental values confirmed the reliability of the QSAR model.
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
2-Amino-5-bromobenzoic acid, 97%
Sigma-Aldrich
1-Naphthol, puriss. p.a., reag. Ph. Eur., ≥99% (GC)
Supelco
Sulfamethizole, VETRANAL®, analytical standard
Supelco
Nitrofurantoin, VETRANAL®, analytical standard
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Supelco
Thiabendazole, PESTANAL®, analytical standard
Supelco
Warfarin, PESTANAL®, analytical standard
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Phenylbutazone
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
1-Naphthol, BioXtra, ≥99%
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Sigma-Aldrich
Thiabendazole, suitable for plant cell culture
Supelco
Sulfamethizole, analytical standard, ≥99% (HPLC)
Sigma-Aldrich
Thiabendazole, ≥99%, powder
Sigma-Aldrich
Dipyridamole, ≥98% (HPLC)
Sigma-Aldrich
Nitrofurantoin, 98.0-102.0% (EP, UV)
Sigma-Aldrich
Sulindac, ≥98.0%
Sigma-Aldrich
Pindolol, ≥98% (TLC), powder