Skip to Content
Merck
All Photos(2)

Key Documents

105910

Sigma-Aldrich

Salicylic acid

ReagentPlus®, ≥99%

Synonym(s):

2-Hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2H
CAS Number:
Molecular Weight:
138.12
Beilstein:
774890
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor density

4.8 (vs air)

Quality Level

vapor pressure

1 mmHg ( 114 °C)

product line

ReagentPlus®

Assay

≥99%

bp

211 °C (lit.)
211 °C/20 mmHg

mp

158-161 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)c1ccccc1O

InChI

1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)

InChI key

YGSDEFSMJLZEOE-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Salicylic acid is an aromatic phenolic derivative. It can be prepared industrially via the Kolbe-Schmitt reaction. It is used as a chemical intermediate in organic synthesis for the synthesis of various organic compounds.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Benfeldt et al.
Acta dermato-venereologica, 79(5), 338-342 (1999-09-24)
Our aim was to simultaneously investigate 2 techniques for in vivo sampling of peripheral compartment pharmacokinetics after systemic administration of acetylsalicylic acid. Ten volunteers were given 2 g acetylsalicylic acid orally. Blood samples and dialysates from 4 microdialysis probes inserted
S Zaugg et al.
Journal of chromatography. B, Biomedical sciences and applications, 752(1), 17-31 (2001-03-20)
Acetylsalicylic acid (Aspirin) is rapidly metabolized to salicylic acid (salicylate) and other compounds, including gentisic acid and salicyluric acid. Monitoring of salicylate and its metabolites is of toxicological, pharmacological and biomedical interest. Three capillary electrophoresis (CE) methods featuring alkaline aqueous
Yunlong Du et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(19), 7946-7951 (2013-04-25)
Removal of cargos from the cell surface via endocytosis is an efficient mechanism to regulate activities of plasma membrane (PM)-resident proteins, such as receptors or transporters. Salicylic acid (SA) is an important plant hormone that is traditionally associated with pathogen
Dorothea Ellinger et al.
Plant physiology, 161(3), 1433-1444 (2013-01-22)
A common response by plants to fungal attack is deposition of callose, a (1,3)-β-glucan polymer, in the form of cell wall thickenings called papillae, at site of wall penetration. While it has been generally believed that the papillae provide a
Loretta Lazzarato et al.
Journal of medicinal chemistry, 51(6), 1894-1903 (2008-02-26)
A new class of products in which the phenol group of salicylic acid is linked to alkanoyl moieties bearing nitrooxy functions has been synthesized and studied for their polyvalent actions. The products were stable in acid and neutral media, while

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service