Merck
  • Multi-target spectral moment QSAR versus ANN for antiparasitic drugs against different parasite species.
Multi-target spectral moment QSAR versus ANN for antiparasitic drugs against different parasite species.
Bioorganic & medicinal chemistry (2010-02-27)
Francisco J Prado-Prado, Xerardo García-Mera, Humberto González-Díaz
ABSTRACT
There are many of pathogen parasite species with different susceptibility profile to antiparasitic drugs. Unfortunately, almost QSAR models predict the biological activity of drugs against only one parasite species. Consequently, predicting the probability with which a drug is active against different species with a single unify model is a goal of the major importance. In so doing, we use Markov Chains theory to calculate new multi-target spectral moments to fit a QSAR model that predict by the first time a mt-QSAR model for 500 drugs tested in the literature against 16 parasite species and other 207 drugs no tested in the literature using spectral moments. The data was processed by linear discriminant analysis (LDA) classifying drugs as active or non-active against the different tested parasite species. The model correctly classifies 311 out of 358 active compounds (86.9%) and 2328 out of 2577 non-active compounds (90.3%) in training series. Overall training performance was 89.9%. Validation of the model was carried out by means of external predicting series. In these series the model classified correctly 157 out 190, 82.6% of antiparasitic compounds and 1151 out of 1277 non-active compounds (90.1%). Overall predictability performance was 89.2%. In addition we developed four types of non Linear Artificial neural networks (ANN) and we compared with the mt-QSAR model. The improved ANN model had an overall training performance was 87%. The present work report the first attempts to calculate within a unify framework probabilities of antiparasitic action of drugs against different parasite species based on spectral moment analysis.
Product Description
Supelco
Betamethasone, VETRANAL®, analytical standard
Supelco
Reserpine Standard for LC-MS, analytical standard, for LC-MS
Supelco
Norfloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Piperine, ≥97%
Sigma-Aldrich
Piperine, ≥95%, FG
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Sigma-Aldrich
Quinine, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
Supelco
Naproxen, VETRANAL®, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
5-Chloro-7-iodo-8-quinolinol, ≥95.0% (HPLC)
Supelco
Sodium decyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
(1S,2S)-(+)-Pseudoephedrine, 98%
Supelco
Ursolic acid, analytical standard
Supelco
Salbutamol, VETRANAL®, analytical standard
Sigma-Aldrich
Quinine, 90%
Sigma-Aldrich
(+)-Cinchonine, 85%
Sigma-Aldrich
Lupeol, ≥94%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Flurbiprofen, cyclooxygenase inhibitor
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Sigma-Aldrich
Reserpine
Supelco
Tolbutamide, analytical standard
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
5,5-Diphenylhydantoin, ≥98%
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Digitoxin, ≥92% (HPLC), powder