Merck
  • Mining biologically-active molecules for inhibitors of fatty acid amide hydrolase (FAAH): identification of phenmedipham and amperozide as FAAH inhibitors.
Mining biologically-active molecules for inhibitors of fatty acid amide hydrolase (FAAH): identification of phenmedipham and amperozide as FAAH inhibitors.
Bioorganic & medicinal chemistry letters (2009-10-24)
Fabien Vincent, Margaret T Nguyen, Daniel E Emerling, Michael G Kelly, Matthew A J Duncton
ABSTRACT
The screening of known medicinal agents against new biological targets has been shown to be a valuable approach for revealing new pharmacology of marketed compounds. Recently, carbamate, urea and ketone inhibitors of fatty acid amide hydrolase (FAAH) have been described as promising treatments for pain, anxiety, depression and other CNS-related conditions. In order to find novel FAAH inhibitors, a focused screen of molecules containing potentially reactive moieties or having in vivo effects that are possibly relevant to the biology of FAAH was conducted. These studies revealed phenmedipham 13 and amperozide 14 to be inhibitors of human FAAH, with an IC(50) of 377 nM and 1.34 microM, respectively.
Product Description
Supelco
Carbofuran, PESTANAL®, analytical standard
Sigma-Aldrich
Fenbendazole, ≥98%
Sigma-Aldrich
Glipizide, solid
Sigma-Aldrich
Propentofylline, solid
Sigma-Aldrich
Streptonigrin from Streptomyces flocculus, ≥98%
Supelco
Propoxur, PESTANAL®, analytical standard
Supelco
Phenmedipham, PESTANAL®, analytical standard
Supelco
Salicylaldehyde, analytical standard
Sigma-Aldrich
Heptanal, ≥95%, FCC, FG
Supelco
Vinclozolin, PESTANAL®, analytical standard
Supelco
Thiophanate-methyl, PESTANAL®, analytical standard
Supelco
Fenbendazole, VETRANAL®, analytical standard
Sigma-Aldrich
Heptaldehyde, 95%
Sigma-Aldrich
Methyl 1-(butylcarbamoyl)-2-benzimidazolecarbamate, 95%
Sigma-Aldrich
Salicylaldehyde, reagent grade, 98%
Sigma-Aldrich
(R)-(+)-Citronellal, technical grade
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
2,4-Dihydroxybenzaldehyde, 98%
Sigma-Aldrich
Carbofuran, 98%
Sigma-Aldrich
Salicylaldehyde, ≥98%, FG
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Supelco
Oxibendazole, VETRANAL®, analytical standard
Supelco
Bendiocarb, PESTANAL®, analytical standard
Supelco
Oxadiazon, PESTANAL®, analytical standard
Supelco
Methiocarb, PESTANAL®, analytical standard
Sigma-Aldrich
Chrysin, ≥96.5%
Sigma-Aldrich
Citral, 95%